Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 95977319-95977566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1I1
Entrez Gene ID
1780
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DNCI1, DNCIC1
UniGene
Hs.440364
Gene Chromosome Location
Chr.7: 95772506-96110322 on Build GRCh38
Transcripts
NM_001135556.1 NM_001135557.1 NM_001278421.1 NM_001278422.1 NM_004411.4 XM_011515861.1 XM_011515862.1 XM_011515863.2 XM_017011804.1 XM_017011805.1 XM_017011806.1 XM_017011807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2865621 c.585G>T p.Q195H Chr.7: 95977555-95977555 on Build GRCh38
748412 c.547G>T p.E183* Chr.7: 95977517-95977517 on Build GRCh38
5830768 c.461C>A p.S154Y Chr.7: 95977542-95977542 on Build GRCh38
5948032 c.444T>C p.D148D Chr.7: 95977525-95977525 on Build GRCh38
1093374 c.572C>A p.S191Y Chr.7: 95977542-95977542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777478919 Chr.7: 95977494-95977495 on Build GRCh38
rs546530856 Chr.7: 95977508-95977508 on Build GRCh38
rs1004575837 Chr.7: 95977513-95977513 on Build GRCh38
rs886221315 Chr.7: 95977406-95977406 on Build GRCh38
rs370031174 Chr.7: 95977539-95977539 on Build GRCh38
rs550248010 Chr.7: 95977454-95977454 on Build GRCh38
rs376286232 Chr.7: 95977489-95977489 on Build GRCh38
rs113015593 Chr.7: 95977404-95977404 on Build GRCh38
rs944992858 Chr.7: 95977425-95977425 on Build GRCh38
rs762550680 Chr.7: 95977515-95977515 on Build GRCh38
rs769156675 Chr.7: 95977464-95977464 on Build GRCh38
rs200097900 Chr.7: 95977485-95977485 on Build GRCh38
rs765771171 Chr.7: 95977533-95977533 on Build GRCh38
rs777195827 Chr.7: 95977468-95977468 on Build GRCh38
rs986115990 Chr.7: 95977346-95977346 on Build GRCh38
rs376550986 Chr.7: 95977476-95977476 on Build GRCh38
rs185161249 Chr.7: 95977481-95977481 on Build GRCh38
rs533685174 Chr.7: 95977501-95977502 on Build GRCh38

View Full Product Details