Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 221429839-221430090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA4
Entrez Gene ID
2043
Gene Name
EPH receptor A4
Gene Aliases
HEK8, SEK, TYRO1
UniGene
Hs.371218
Gene Chromosome Location
Chr.2: 221418027-221574202 on Build GRCh38
Transcripts
NM_001304536.1 NM_001304537.1 NM_004438.4 XM_005246374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414707 c.2648G>A p.R883H Chr.2: 221430000-221430000 on Build GRCh38
1016963 c.2571G>A p.M857I Chr.2: 221430077-221430077 on Build GRCh38
4790016 c.2595G>C p.R865S Chr.2: 221430053-221430053 on Build GRCh38
7197367 c.2609A>C p.K870T Chr.2: 221430039-221430039 on Build GRCh38
1016961 c.2600A>G p.D867G Chr.2: 221430048-221430048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762833404 Chr.2: 221429952-221429952 on Build GRCh38
rs374390918 Chr.2: 221429979-221429979 on Build GRCh38
rs974671130 Chr.2: 221429891-221429891 on Build GRCh38
rs1034234174 Chr.2: 221430014-221430014 on Build GRCh38
rs1002132191 Chr.2: 221429990-221429990 on Build GRCh38
rs771148130 Chr.2: 221430060-221430060 on Build GRCh38
rs776407691 Chr.2: 221429927-221429927 on Build GRCh38
rs776926049 Chr.2: 221430070-221430070 on Build GRCh38
rs765523420 Chr.2: 221430000-221430000 on Build GRCh38
rs371830529 Chr.2: 221429969-221429969 on Build GRCh38
rs767587519 Chr.2: 221429964-221429964 on Build GRCh38
rs886484557 Chr.2: 221429955-221429955 on Build GRCh38
rs374568057 Chr.2: 221429917-221429917 on Build GRCh38
rs760466873 Chr.2: 221429963-221429963 on Build GRCh38
rs777177020 Chr.2: 221429909-221429909 on Build GRCh38
rs780769305 Chr.2: 221430043-221430043 on Build GRCh38
rs755335356 Chr.2: 221429994-221429994 on Build GRCh38
rs745528063 Chr.2: 221430049-221430049 on Build GRCh38
rs930615257 Chr.2: 221429948-221429948 on Build GRCh38
rs754240886 Chr.2: 221429982-221429982 on Build GRCh38
rs201216689 Chr.2: 221429932-221429932 on Build GRCh38
rs768512485 Chr.2: 221430056-221430056 on Build GRCh38
rs142296640 Chr.2: 221430001-221430001 on Build GRCh38
rs146939684 Chr.2: 221429972-221429972 on Build GRCh38
rs746643824 Chr.2: 221429913-221429913 on Build GRCh38
rs749224993 Chr.2: 221430051-221430051 on Build GRCh38
rs868613699 Chr.2: 221430033-221430033 on Build GRCh38
rs773418289 Chr.2: 221430057-221430057 on Build GRCh38
rs779738045 Chr.2: 221430050-221430050 on Build GRCh38
rs769493680 Chr.2: 221429934-221429934 on Build GRCh38
rs1049067360 Chr.2: 221429950-221429950 on Build GRCh38
rs770611286 Chr.2: 221429921-221429921 on Build GRCh38
rs142860268 Chr.2: 221429970-221429970 on Build GRCh38
rs760891696 Chr.2: 221430059-221430059 on Build GRCh38
rs563009251 Chr.2: 221429907-221429907 on Build GRCh38
rs188379183 Chr.2: 221429940-221429940 on Build GRCh38
rs758880801 Chr.2: 221430002-221430002 on Build GRCh38
rs751138156 Chr.2: 221430020-221430020 on Build GRCh38
rs777306913 Chr.2: 221430007-221430007 on Build GRCh38
rs756889351 Chr.2: 221430036-221430036 on Build GRCh38

View Full Product Details