Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116278547-116278806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSE
Entrez Gene ID
29940
Gene Name
dermatan sulfate epimerase
Gene Aliases
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
UniGene
Hs.458358
Gene Chromosome Location
Chr.6: 116254152-116441261 on Build GRCh38
Transcripts
NM_001322937.1 NM_001322938.1 NM_001322940.1 NM_003309.3

Gene Symbol
TSPYL1
Entrez Gene ID
7259
Gene Name
TSPY like 1
Gene Aliases
TSPYL
UniGene
Hs.458358
Gene Chromosome Location
Chr.6: 116274859-116280117 on Build GRCh38
Transcripts
NM_001322937.1 NM_001322938.1 NM_001322940.1 NM_003309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1329351 c.1057A>T p.T353S Chr.6: 116278774-116278774 on Build GRCh38
1439967 c.1258C>T p.R420C Chr.6: 116278573-116278573 on Build GRCh38
6038974 c.1231C>T p.R411C Chr.6: 116278600-116278600 on Build GRCh38
6810694 c.1103G>A p.R368H Chr.6: 116278728-116278728 on Build GRCh38
3828863 c.1220A>G p.Y407C Chr.6: 116278611-116278611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754392904 Chr.6: 116278629-116278629 on Build GRCh38
rs769775172 Chr.6: 116278787-116278787 on Build GRCh38
rs756171541 Chr.6: 116278589-116278589 on Build GRCh38
rs748090346 Chr.6: 116278784-116278784 on Build GRCh38
rs979950673 Chr.6: 116278759-116278759 on Build GRCh38
rs764097142 Chr.6: 116278731-116278731 on Build GRCh38
rs768196964 Chr.6: 116278772-116278772 on Build GRCh38
rs779485039 Chr.6: 116278647-116278647 on Build GRCh38
rs948915024 Chr.6: 116278601-116278601 on Build GRCh38
rs750767335 Chr.6: 116278754-116278754 on Build GRCh38
rs45490498 Chr.6: 116278696-116278696 on Build GRCh38
rs140756663 Chr.6: 116278733-116278733 on Build GRCh38
rs965025042 Chr.6: 116278776-116278776 on Build GRCh38
rs376103333 Chr.6: 116278581-116278581 on Build GRCh38
rs760742976 Chr.6: 116278729-116278729 on Build GRCh38
rs781703026 Chr.6: 116278733-116278735 on Build GRCh38
rs751770322 Chr.6: 116278572-116278572 on Build GRCh38
rs755262799 Chr.6: 116278573-116278573 on Build GRCh38
rs1044994210 Chr.6: 116278613-116278613 on Build GRCh38
rs746979549 Chr.6: 116278637-116278637 on Build GRCh38
rs767682435 Chr.6: 116278719-116278719 on Build GRCh38
rs749764449 Chr.6: 116278623-116278623 on Build GRCh38
rs200556782 Chr.6: 116278742-116278742 on Build GRCh38
rs758865548 Chr.6: 116278757-116278757 on Build GRCh38
rs371415913 Chr.6: 116278718-116278718 on Build GRCh38
rs537676463 Chr.6: 116278599-116278599 on Build GRCh38
rs549729790 Chr.6: 116278672-116278672 on Build GRCh38
rs761711631 Chr.6: 116278698-116278698 on Build GRCh38
rs745484190 Chr.6: 116278650-116278650 on Build GRCh38
rs369165300 Chr.6: 116278587-116278587 on Build GRCh38
rs752948355 Chr.6: 116278727-116278727 on Build GRCh38
rs746483626 Chr.6: 116278678-116278678 on Build GRCh38
rs144676525 Chr.6: 116278764-116278764 on Build GRCh38
rs778304109 Chr.6: 116278591-116278591 on Build GRCh38
rs752827756 Chr.6: 116278584-116278584 on Build GRCh38
rs377679883 Chr.6: 116278736-116278736 on Build GRCh38
rs779397957 Chr.6: 116278744-116278744 on Build GRCh38
rs996015480 Chr.6: 116278786-116278786 on Build GRCh38
rs768052382 Chr.6: 116278686-116278686 on Build GRCh38
rs370128037 Chr.6: 116278700-116278700 on Build GRCh38
rs746652096 Chr.6: 116278765-116278765 on Build GRCh38
rs773035028 Chr.6: 116278709-116278709 on Build GRCh38
rs758204216 Chr.6: 116278575-116278575 on Build GRCh38
rs373244327 Chr.6: 116278778-116278778 on Build GRCh38
rs764951902 Chr.6: 116278571-116278571 on Build GRCh38
rs773159856 Chr.6: 116278788-116278788 on Build GRCh38
rs777781994 Chr.6: 116278592-116278592 on Build GRCh38
rs889535498 Chr.6: 116278631-116278631 on Build GRCh38
rs771898385 Chr.6: 116278659-116278659 on Build GRCh38

View Full Product Details