Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168896450-168896695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR4
Entrez Gene ID
84869
Gene Name
carbonyl reductase 4
Gene Aliases
SDR45C1
UniGene
Hs.659311
Gene Chromosome Location
Chr.4: 168894484-169010317 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Gene Symbol
PALLD
Entrez Gene ID
23022
Gene Name
palladin, cytoskeletal associated protein
Gene Aliases
CGI-151, CGI151, MYN, PNCA1, SIH002
UniGene
Hs.151220
Gene Chromosome Location
Chr.4: 168497039-168928457 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

SNP Detail

SNP ID Chromosome Location
rs749252340 Chr.4: 168896600-168896600 on Build GRCh38
rs766416032 Chr.4: 168896530-168896530 on Build GRCh38
rs928746600 Chr.4: 168896547-168896547 on Build GRCh38
rs982847011 Chr.4: 168896526-168896526 on Build GRCh38
rs760412674 Chr.4: 168896489-168896489 on Build GRCh38
rs780011911 Chr.4: 168896638-168896638 on Build GRCh38
rs915812176 Chr.4: 168896646-168896646 on Build GRCh38
rs373210861 Chr.4: 168896668-168896668 on Build GRCh38
rs571138706 Chr.4: 168896534-168896534 on Build GRCh38
rs1015502212 Chr.4: 168896545-168896545 on Build GRCh38
rs756024689 Chr.4: 168896574-168896574 on Build GRCh38
rs1014056261 Chr.4: 168896664-168896664 on Build GRCh38
rs991371952 Chr.4: 168896588-168896588 on Build GRCh38
rs1004576504 Chr.4: 168896496-168896496 on Build GRCh38
rs538974015 Chr.4: 168896628-168896628 on Build GRCh38
rs947328360 Chr.4: 168896655-168896655 on Build GRCh38
rs781098627 Chr.4: 168896504-168896504 on Build GRCh38
rs539957451 Chr.4: 168896595-168896595 on Build GRCh38
rs951437937 Chr.4: 168896506-168896506 on Build GRCh38

View Full Product Details