Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11423922-11424151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC151
Entrez Gene ID
115948
Gene Name
coiled-coil domain containing 151
Gene Aliases
CILD30
UniGene
Hs.124010
Gene Chromosome Location
Chr.19: 11420605-11435782 on Build GRCh38
Transcripts
NM_001302453.1 NM_001302454.1 NM_145045.4 XM_017026241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4074170 c.970C>T p.R324C Chr.19: 11424023-11424023 on Build GRCh38
7156298 c.987A>G p.L329L Chr.19: 11424006-11424006 on Build GRCh38
1680672 c.1045C>A p.R349S Chr.19: 11423948-11423948 on Build GRCh38
6299419 c.1043A>C p.Q348P Chr.19: 11423950-11423950 on Build GRCh38
1199887 c.995A>T p.D332V Chr.19: 11423998-11423998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756771521 Chr.19: 11424123-11424123 on Build GRCh38
rs112797378 Chr.19: 11423996-11423996 on Build GRCh38
rs938305782 Chr.19: 11424122-11424122 on Build GRCh38
rs200295181 Chr.19: 11424058-11424058 on Build GRCh38
rs1026820959 Chr.19: 11424091-11424091 on Build GRCh38
rs568968215 Chr.19: 11424129-11424129 on Build GRCh38
rs778052460 Chr.19: 11424032-11424032 on Build GRCh38
rs367780527 Chr.19: 11424118-11424118 on Build GRCh38
rs539181030 Chr.19: 11424130-11424130 on Build GRCh38
rs746096719 Chr.19: 11423994-11423996 on Build GRCh38
rs767596988 Chr.19: 11423979-11423979 on Build GRCh38
rs764817042 Chr.19: 11424073-11424073 on Build GRCh38
rs546923726 Chr.19: 11424017-11424017 on Build GRCh38
rs749889069 Chr.19: 11424075-11424075 on Build GRCh38
rs888388277 Chr.19: 11424098-11424098 on Build GRCh38
rs756919168 Chr.19: 11424000-11424000 on Build GRCh38
rs749500188 Chr.19: 11424041-11424041 on Build GRCh38
rs930790607 Chr.19: 11424092-11424092 on Build GRCh38
rs780372852 Chr.19: 11424063-11424064 on Build GRCh38
rs749831964 Chr.19: 11424067-11424068 on Build GRCh38
rs770110931 Chr.19: 11424050-11424051 on Build GRCh38
rs774452185 Chr.19: 11424048-11424048 on Build GRCh38
rs1039132115 Chr.19: 11424061-11424061 on Build GRCh38
rs774347087 Chr.19: 11423961-11423961 on Build GRCh38
rs772078230 Chr.19: 11424051-11424051 on Build GRCh38
rs1053136709 Chr.19: 11423958-11423958 on Build GRCh38
rs769942319 Chr.19: 11423952-11423952 on Build GRCh38
rs773425892 Chr.19: 11423953-11423953 on Build GRCh38
rs749932786 Chr.19: 11424010-11424010 on Build GRCh38
rs764772748 Chr.19: 11424004-11424004 on Build GRCh38
rs371436780 Chr.19: 11424050-11424050 on Build GRCh38
rs775379203 Chr.19: 11423989-11423989 on Build GRCh38
rs763147096 Chr.19: 11423956-11423956 on Build GRCh38
rs751315764 Chr.19: 11424090-11424090 on Build GRCh38
rs368616907 Chr.19: 11423997-11423997 on Build GRCh38
rs781672649 Chr.19: 11424021-11424021 on Build GRCh38
rs748442231 Chr.19: 11424023-11424023 on Build GRCh38
rs899667975 Chr.19: 11424078-11424078 on Build GRCh38
rs559900157 Chr.19: 11424045-11424045 on Build GRCh38
rs763968197 Chr.19: 11424059-11424059 on Build GRCh38
rs370746583 Chr.19: 11423964-11423964 on Build GRCh38
rs967633646 Chr.19: 11423991-11423991 on Build GRCh38
rs905437426 Chr.19: 11424114-11424114 on Build GRCh38
rs771026924 Chr.19: 11423957-11423957 on Build GRCh38
rs550393980 Chr.19: 11424068-11424068 on Build GRCh38
rs776329735 Chr.19: 11424066-11424066 on Build GRCh38

View Full Product Details