Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 113001212-113001471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP2
Entrez Gene ID
167227
Gene Name
decapping mRNA 2
Gene Aliases
NUDT20
UniGene
Hs.443875
Gene Chromosome Location
Chr.5: 112976702-113022195 on Build GRCh38
Transcripts
NM_001242377.1 NM_152624.5 NR_038352.1 XM_017009163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005938 c.668A>G p.N223S Chr.5: 113001439-113001439 on Build GRCh38
4676574 c.672A>T p.K224N Chr.5: 113001443-113001443 on Build GRCh38
1203152 c.671A>C p.K224T Chr.5: 113001442-113001442 on Build GRCh38
5816044 c.652C>A p.L218I Chr.5: 113001423-113001423 on Build GRCh38
4552927 c.575G>A p.R192K Chr.5: 113001226-113001226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902457072 Chr.5: 113001316-113001316 on Build GRCh38
rs760057225 Chr.5: 113001373-113001373 on Build GRCh38
rs758099700 Chr.5: 113001284-113001284 on Build GRCh38
rs760803146 Chr.5: 113001258-113001258 on Build GRCh38
rs112761672 Chr.5: 113001408-113001408 on Build GRCh38
rs749134044 Chr.5: 113001335-113001335 on Build GRCh38
rs756878494 Chr.5: 113001406-113001406 on Build GRCh38
rs889639685 Chr.5: 113001328-113001328 on Build GRCh38
rs780026913 Chr.5: 113001433-113001433 on Build GRCh38
rs764554877 Chr.5: 113001393-113001393 on Build GRCh38
rs376632643 Chr.5: 113001371-113001371 on Build GRCh38
rs901561034 Chr.5: 113001443-113001443 on Build GRCh38
rs756058426 Chr.5: 113001420-113001420 on Build GRCh38
rs756717478 Chr.5: 113001309-113001309 on Build GRCh38
rs1019523383 Chr.5: 113001436-113001436 on Build GRCh38
rs780717459 Chr.5: 113001413-113001413 on Build GRCh38
rs766793855 Chr.5: 113001268-113001268 on Build GRCh38
rs765710158 Chr.5: 113001375-113001375 on Build GRCh38
rs758896820 Chr.5: 113001392-113001392 on Build GRCh38
rs766424495 Chr.5: 113001358-113001358 on Build GRCh38
rs745414532 Chr.5: 113001419-113001419 on Build GRCh38
rs751195051 Chr.5: 113001401-113001401 on Build GRCh38
rs781065228 Chr.5: 113001383-113001383 on Build GRCh38
rs772270550 Chr.5: 113001337-113001337 on Build GRCh38
rs370145343 Chr.5: 113001310-113001310 on Build GRCh38
rs753096232 Chr.5: 113001376-113001376 on Build GRCh38
rs765596511 Chr.5: 113001279-113001279 on Build GRCh38
rs769653680 Chr.5: 113001318-113001318 on Build GRCh38
rs760892590 Chr.5: 113001341-113001341 on Build GRCh38
rs745766156 Chr.5: 113001311-113001311 on Build GRCh38
rs202191634 Chr.5: 113001308-113001308 on Build GRCh38
rs115165195 Chr.5: 113001283-113001283 on Build GRCh38
rs373545100 Chr.5: 113001329-113001329 on Build GRCh38
rs369984345 Chr.5: 113001442-113001442 on Build GRCh38
rs775238310 Chr.5: 113001331-113001331 on Build GRCh38
rs934983913 Chr.5: 113001386-113001386 on Build GRCh38
rs1006675140 Chr.5: 113001414-113001414 on Build GRCh38
rs755455187 Chr.5: 113001277-113001277 on Build GRCh38
rs754305101 Chr.5: 113001269-113001269 on Build GRCh38
rs1043109801 Chr.5: 113001290-113001290 on Build GRCh38
rs754818941 Chr.5: 113001246-113001247 on Build GRCh38
rs777625022 Chr.5: 113001288-113001288 on Build GRCh38

View Full Product Details