Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113415466-113415693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
UniGene
Hs.73893
Gene Chromosome Location
Chr.11: 113409595-113475279 on Build GRCh38
Transcripts
NM_000795.3 NM_016574.3 XM_017017296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443548 c.633G>A p.K211K Chr.11: 113415511-113415511 on Build GRCh38
3443552 c.573C>T p.V191V Chr.11: 113415571-113415571 on Build GRCh38
6579346 c.557delA p.N186fs*51 Chr.11: 113415587-113415587 on Build GRCh38
3443545 c.675C>T p.T225T Chr.11: 113415469-113415469 on Build GRCh38
4017916 c.649C>T p.R217C Chr.11: 113415495-113415495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185801473 Chr.11: 113415648-113415648 on Build GRCh38
rs781572485 Chr.11: 113415516-113415516 on Build GRCh38
rs1043241299 Chr.11: 113415618-113415618 on Build GRCh38
rs143890565 Chr.11: 113415610-113415610 on Build GRCh38
rs1039382986 Chr.11: 113415641-113415641 on Build GRCh38
rs1057032656 Chr.11: 113415608-113415608 on Build GRCh38
rs775101965 Chr.11: 113415583-113415583 on Build GRCh38
rs201451627 Chr.11: 113415617-113415617 on Build GRCh38
rs997751506 Chr.11: 113415654-113415654 on Build GRCh38
rs866197752 Chr.11: 113415611-113415611 on Build GRCh38
rs761671516 Chr.11: 113415614-113415614 on Build GRCh38
rs755559594 Chr.11: 113415486-113415486 on Build GRCh38
rs200148328 Chr.11: 113415488-113415488 on Build GRCh38
rs557036496 Chr.11: 113415485-113415485 on Build GRCh38
rs1030596917 Chr.11: 113415656-113415656 on Build GRCh38
rs201949556 Chr.11: 113415629-113415629 on Build GRCh38
rs771849191 Chr.11: 113415577-113415577 on Build GRCh38
rs573022587 Chr.11: 113415649-113415649 on Build GRCh38
rs904287542 Chr.11: 113415622-113415622 on Build GRCh38
rs758685318 Chr.11: 113415624-113415624 on Build GRCh38
rs202200970 Chr.11: 113415565-113415565 on Build GRCh38
rs199731353 Chr.11: 113415657-113415657 on Build GRCh38
rs987305741 Chr.11: 113415489-113415489 on Build GRCh38
rs866976053 Chr.11: 113415551-113415551 on Build GRCh38
rs200496289 Chr.11: 113415604-113415604 on Build GRCh38
rs778351726 Chr.11: 113415662-113415662 on Build GRCh38
rs202133725 Chr.11: 113415615-113415615 on Build GRCh38
rs147086003 Chr.11: 113415526-113415526 on Build GRCh38
rs773657682 Chr.11: 113415559-113415559 on Build GRCh38
rs961789208 Chr.11: 113415571-113415571 on Build GRCh38
rs373761235 Chr.11: 113415556-113415556 on Build GRCh38
rs938621123 Chr.11: 113415597-113415597 on Build GRCh38
rs913088665 Chr.11: 113415616-113415616 on Build GRCh38
rs138509142 Chr.11: 113415546-113415546 on Build GRCh38
rs200267533 Chr.11: 113415630-113415630 on Build GRCh38

View Full Product Details