Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137050777-137051038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3655935 c.721C>T p.L241F Chr.9: 137050955-137050955 on Build GRCh38
239709 c.716_724delGCTTCCTCT p.F240_C242delFLC Chr.9: 137050952-137050960 on Build GRCh38
1739559 c.733C>T p.R245C Chr.9: 137050943-137050943 on Build GRCh38
351865 c.739C>T p.Q247* Chr.9: 137050937-137050937 on Build GRCh38
422431 c.747C>G p.L249L Chr.9: 137050929-137050929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141307913 Chr.9: 137050988-137050988 on Build GRCh38
rs2292925 Chr.9: 137050911-137050911 on Build GRCh38
rs116635795 Chr.9: 137050962-137050962 on Build GRCh38
rs199705392 Chr.9: 137050968-137050968 on Build GRCh38
rs770820178 Chr.9: 137050938-137050938 on Build GRCh38
rs761081418 Chr.9: 137050977-137050977 on Build GRCh38
rs781245560 Chr.9: 137050975-137050976 on Build GRCh38
rs773510818 Chr.9: 137050976-137050976 on Build GRCh38
rs879376155 Chr.9: 137050888-137050888 on Build GRCh38
rs768876583 Chr.9: 137050954-137050954 on Build GRCh38
rs368559678 Chr.9: 137050863-137050863 on Build GRCh38
rs756649518 Chr.9: 137050902-137050902 on Build GRCh38
rs761409849 Chr.9: 137050915-137050915 on Build GRCh38
rs767845036 Chr.9: 137050975-137050975 on Build GRCh38
rs768004591 Chr.9: 137050898-137050898 on Build GRCh38
rs759351054 Chr.9: 137050943-137050943 on Build GRCh38
rs750747096 Chr.9: 137050901-137050901 on Build GRCh38
rs757106831 Chr.9: 137051010-137051010 on Build GRCh38
rs541605947 Chr.9: 137051000-137051000 on Build GRCh38
rs780800770 Chr.9: 137051011-137051011 on Build GRCh38
rs927313534 Chr.9: 137050822-137050822 on Build GRCh38
rs374065956 Chr.9: 137050942-137050942 on Build GRCh38
rs760544909 Chr.9: 137050855-137050855 on Build GRCh38
rs777549961 Chr.9: 137050924-137050924 on Build GRCh38
rs763064182 Chr.9: 137050877-137050877 on Build GRCh38
rs993142652 Chr.9: 137050847-137050847 on Build GRCh38
rs770836776 Chr.9: 137050861-137050861 on Build GRCh38
rs747820624 Chr.9: 137050914-137050914 on Build GRCh38
rs769190505 Chr.9: 137050982-137050982 on Build GRCh38
rs753341928 Chr.9: 137050908-137050908 on Build GRCh38
rs367870548 Chr.9: 137051009-137051009 on Build GRCh38
rs746804655 Chr.9: 137050932-137050932 on Build GRCh38
rs965890788 Chr.9: 137050960-137050960 on Build GRCh38
rs764450531 Chr.9: 137050884-137050884 on Build GRCh38
rs780600887 Chr.9: 137050905-137050905 on Build GRCh38
rs145240352 Chr.9: 137050998-137050998 on Build GRCh38
rs201122803 Chr.9: 137050989-137050989 on Build GRCh38
rs535221430 Chr.9: 137050890-137050890 on Build GRCh38
rs777380815 Chr.9: 137051005-137051005 on Build GRCh38
rs771657320 Chr.9: 137050922-137050922 on Build GRCh38
rs775570161 Chr.9: 137050910-137050910 on Build GRCh38

View Full Product Details