Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105008598-105008867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP5
Entrez Gene ID
838
Gene Name
caspase 5
Gene Aliases
ICE(rel)III, ICEREL-III, ICH-3
UniGene
Hs.213327
Gene Chromosome Location
Chr.11: 104994240-105023902 on Build GRCh38
Transcripts
NM_001136109.1 NM_001136110.1 NM_001136112.1 NM_004347.3 NR_024239.1 NR_036562.1 XM_011543021.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585299 c.194C>T p.T65M Chr.11: 105008833-105008833 on Build GRCh38
1704513 c.85A>G p.I29V Chr.11: 105008855-105008855 on Build GRCh38
4017424 c.206C>T p.S69L Chr.11: 105008821-105008821 on Build GRCh38
1704512 c.172A>G p.I58V Chr.11: 105008855-105008855 on Build GRCh38
4017425 c.119C>T p.S40L Chr.11: 105008821-105008821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777253451 Chr.11: 105008807-105008807 on Build GRCh38
rs150404959 Chr.11: 105008815-105008815 on Build GRCh38
rs774406316 Chr.11: 105008723-105008723 on Build GRCh38
rs2276414 Chr.11: 105008835-105008835 on Build GRCh38
rs773974557 Chr.11: 105008814-105008814 on Build GRCh38
rs755946098 Chr.11: 105008822-105008822 on Build GRCh38
rs772262415 Chr.11: 105008781-105008781 on Build GRCh38
rs770297060 Chr.11: 105008776-105008777 on Build GRCh38
rs185729600 Chr.11: 105008832-105008832 on Build GRCh38
rs1013856883 Chr.11: 105008661-105008661 on Build GRCh38
rs200310283 Chr.11: 105008840-105008840 on Build GRCh38
rs749543933 Chr.11: 105008628-105008628 on Build GRCh38
rs181553706 Chr.11: 105008620-105008620 on Build GRCh38
rs777767473 Chr.11: 105008762-105008762 on Build GRCh38
rs922976355 Chr.11: 105008619-105008619 on Build GRCh38
rs372557464 Chr.11: 105008636-105008636 on Build GRCh38
rs958156742 Chr.11: 105008751-105008751 on Build GRCh38
rs963151086 Chr.11: 105008687-105008687 on Build GRCh38
rs762222274 Chr.11: 105008808-105008808 on Build GRCh38
rs1036555214 Chr.11: 105008647-105008647 on Build GRCh38
rs1002583726 Chr.11: 105008724-105008724 on Build GRCh38
rs201768775 Chr.11: 105008645-105008645 on Build GRCh38
rs201179331 Chr.11: 105008833-105008833 on Build GRCh38
rs1020104761 Chr.11: 105008783-105008783 on Build GRCh38
rs752221620 Chr.11: 105008820-105008820 on Build GRCh38
rs768837150 Chr.11: 105008805-105008805 on Build GRCh38
rs747432616 Chr.11: 105008789-105008789 on Build GRCh38
rs766026230 Chr.11: 105008809-105008809 on Build GRCh38
rs778963224 Chr.11: 105008773-105008773 on Build GRCh38
rs746132369 Chr.11: 105008775-105008775 on Build GRCh38
rs967155921 Chr.11: 105008784-105008784 on Build GRCh38
rs376017084 Chr.11: 105008764-105008764 on Build GRCh38
rs771060633 Chr.11: 105008772-105008772 on Build GRCh38
rs948741955 Chr.11: 105008785-105008785 on Build GRCh38
rs1015918288 Chr.11: 105008640-105008640 on Build GRCh38
rs566215693 Chr.11: 105008625-105008625 on Build GRCh38
rs771233663 Chr.11: 105008630-105008630 on Build GRCh38
rs905192757 Chr.11: 105008688-105008688 on Build GRCh38
rs1025622718 Chr.11: 105008667-105008667 on Build GRCh38
rs767306878 Chr.11: 105008819-105008819 on Build GRCh38
rs45516397 Chr.11: 105008747-105008747 on Build GRCh38
rs551852161 Chr.11: 105008654-105008654 on Build GRCh38
rs868333019 Chr.11: 105008701-105008701 on Build GRCh38

View Full Product Details