Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42521146-42521419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM1
Entrez Gene ID
634
Gene Name
carcinoembryonic antigen related cell adhesion molecule 1
Gene Aliases
BGP, BGP1, BGPI
UniGene
Hs.512682
Gene Chromosome Location
Chr.19: 42507306-42528509 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 XM_011527206.1 NR_073179.1 NR_073180.1

Gene Symbol
LIPE-AS1
Entrez Gene ID
100996307
Gene Name
LIPE antisense RNA 1
Gene Aliases
-
UniGene
Hs.466840
Gene Chromosome Location
Chr.19: 42397148-42652355 on Build GRCh38
Transcripts
NM_001024912.2 NM_001184813.1 NM_001184815.1 NM_001184816.1 NM_001205344.1 NM_001712.4 XM_011527206.1 NR_073179.1 NR_073180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4796810 c.923G>A p.C308Y Chr.19: 42521302-42521302 on Build GRCh38
239284 c.893C>T p.T298I Chr.19: 42521332-42521332 on Build GRCh38
4745741 c.906T>C p.N302N Chr.19: 42521319-42521319 on Build GRCh38
4127682 c.958+3A>C p.? Chr.19: 42521264-42521264 on Build GRCh38
6625881 c.814C>T p.L272F Chr.19: 42521411-42521411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760040548 Chr.19: 42521240-42521240 on Build GRCh38
rs779826417 Chr.19: 42521259-42521262 on Build GRCh38
rs751596811 Chr.19: 42521233-42521233 on Build GRCh38
rs760674584 Chr.19: 42521388-42521388 on Build GRCh38
rs768483408 Chr.19: 42521349-42521350 on Build GRCh38
rs146410449 Chr.19: 42521279-42521279 on Build GRCh38
rs950475073 Chr.19: 42521172-42521172 on Build GRCh38
rs770117311 Chr.19: 42521214-42521214 on Build GRCh38
rs772941194 Chr.19: 42521324-42521324 on Build GRCh38
rs745830665 Chr.19: 42521228-42521229 on Build GRCh38
rs983374585 Chr.19: 42521182-42521182 on Build GRCh38
rs756402877 Chr.19: 42521277-42521277 on Build GRCh38
rs774427095 Chr.19: 42521356-42521356 on Build GRCh38
rs764400411 Chr.19: 42521389-42521389 on Build GRCh38
rs763067440 Chr.19: 42521222-42521222 on Build GRCh38
rs140710372 Chr.19: 42521280-42521280 on Build GRCh38
rs548335226 Chr.19: 42521188-42521188 on Build GRCh38
rs768693621 Chr.19: 42521289-42521289 on Build GRCh38
rs1027326866 Chr.19: 42521171-42521171 on Build GRCh38
rs746464510 Chr.19: 42521284-42521284 on Build GRCh38
rs868638243 Chr.19: 42521382-42521382 on Build GRCh38
rs766467980 Chr.19: 42521230-42521230 on Build GRCh38
rs991840343 Chr.19: 42521288-42521288 on Build GRCh38
rs749144302 Chr.19: 42521333-42521336 on Build GRCh38
rs752144585 Chr.19: 42521274-42521275 on Build GRCh38
rs753124950 Chr.19: 42521383-42521383 on Build GRCh38
rs764537360 Chr.19: 42521231-42521231 on Build GRCh38
rs755984290 Chr.19: 42521230-42521239 on Build GRCh38
rs767853979 Chr.19: 42521261-42521261 on Build GRCh38
rs768109330 Chr.19: 42521372-42521372 on Build GRCh38
rs748012689 Chr.19: 42521302-42521302 on Build GRCh38
rs761196092 Chr.19: 42521385-42521385 on Build GRCh38
rs780982926 Chr.19: 42521301-42521301 on Build GRCh38
rs561744466 Chr.19: 42521216-42521216 on Build GRCh38
rs763190977 Chr.19: 42521333-42521333 on Build GRCh38
rs759609108 Chr.19: 42521361-42521361 on Build GRCh38
rs148859595 Chr.19: 42521369-42521369 on Build GRCh38
rs750261660 Chr.19: 42521185-42521185 on Build GRCh38
rs550461015 Chr.19: 42521337-42521337 on Build GRCh38
rs376242453 Chr.19: 42521316-42521316 on Build GRCh38
rs1024665085 Chr.19: 42521266-42521266 on Build GRCh38
rs576921764 Chr.19: 42521198-42521198 on Build GRCh38
rs971230263 Chr.19: 42521275-42521275 on Build GRCh38
rs753172863 Chr.19: 42521268-42521268 on Build GRCh38
rs150099950 Chr.19: 42521281-42521281 on Build GRCh38

View Full Product Details