Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40941160-40941407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELF1
Entrez Gene ID
1997
Gene Name
E74 like ETS transcription factor 1
Gene Aliases
-
UniGene
Hs.135646
Gene Chromosome Location
Chr.13: 40931919-41061414 on Build GRCh38
Transcripts
NM_001145353.1 NM_172373.3 XM_005266276.1 XM_005266277.3 XM_011534950.1 XM_011534951.2 XM_017020409.1 XM_017020410.1 XM_017020411.1 XM_017020412.1 XM_017020413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
389099 c.807-1_807GG>TC p.? Chr.13: 40941370-40941371 on Build GRCh38
4455615 c.835A>T p.K279* Chr.13: 40941342-40941342 on Build GRCh38
5000215 c.851G>T p.R284L Chr.13: 40941326-40941326 on Build GRCh38
5568104 c.1002T>A p.S334R Chr.13: 40941175-40941175 on Build GRCh38
1747441 c.962C>T p.S321L Chr.13: 40941215-40941215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7987185 Chr.13: 40941235-40941235 on Build GRCh38
rs770939173 Chr.13: 40941222-40941222 on Build GRCh38
rs140646954 Chr.13: 40941194-40941194 on Build GRCh38
rs777768092 Chr.13: 40941213-40941213 on Build GRCh38
rs1009748953 Chr.13: 40941319-40941319 on Build GRCh38
rs762084486 Chr.13: 40941238-40941238 on Build GRCh38
rs778527444 Chr.13: 40941206-40941206 on Build GRCh38
rs144532940 Chr.13: 40941209-40941209 on Build GRCh38
rs532698900 Chr.13: 40941252-40941252 on Build GRCh38
rs750744628 Chr.13: 40941191-40941191 on Build GRCh38
rs754470942 Chr.13: 40941205-40941205 on Build GRCh38
rs757021667 Chr.13: 40941324-40941324 on Build GRCh38
rs373079831 Chr.13: 40941264-40941264 on Build GRCh38
rs371166882 Chr.13: 40941358-40941358 on Build GRCh38
rs777535025 Chr.13: 40941297-40941297 on Build GRCh38
rs201199410 Chr.13: 40941325-40941325 on Build GRCh38
rs76461227 Chr.13: 40941215-40941215 on Build GRCh38
rs753438585 Chr.13: 40941201-40941201 on Build GRCh38
rs774343467 Chr.13: 40941236-40941236 on Build GRCh38
rs142074159 Chr.13: 40941361-40941361 on Build GRCh38
rs755476257 Chr.13: 40941254-40941254 on Build GRCh38
rs754334265 Chr.13: 40941253-40941253 on Build GRCh38
rs1032880556 Chr.13: 40941286-40941286 on Build GRCh38
rs370523749 Chr.13: 40941218-40941218 on Build GRCh38
rs767868893 Chr.13: 40941190-40941190 on Build GRCh38
rs760268434 Chr.13: 40941279-40941280 on Build GRCh38
rs773668917 Chr.13: 40941240-40941240 on Build GRCh38
rs143469326 Chr.13: 40941291-40941291 on Build GRCh38
rs202087877 Chr.13: 40941282-40941282 on Build GRCh38
rs758066781 Chr.13: 40941211-40941211 on Build GRCh38
rs776730565 Chr.13: 40941354-40941354 on Build GRCh38
rs146850967 Chr.13: 40941188-40941188 on Build GRCh38
rs767725443 Chr.13: 40941239-40941239 on Build GRCh38
rs137863135 Chr.13: 40941330-40941330 on Build GRCh38
rs766771438 Chr.13: 40941195-40941195 on Build GRCh38
rs746867928 Chr.13: 40941298-40941298 on Build GRCh38
rs9562248 Chr.13: 40941232-40941232 on Build GRCh38
rs990847340 Chr.13: 40941186-40941186 on Build GRCh38
rs745794216 Chr.13: 40941345-40941345 on Build GRCh38
rs12876555 Chr.13: 40941331-40941331 on Build GRCh38

View Full Product Details