Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36930552-36930817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636117 c.866G>T p.C289F Chr.22: 36930684-36930684 on Build GRCh38
6526520 c.870C>A p.S290S Chr.22: 36930688-36930688 on Build GRCh38
478930 c.965C>G p.S322C Chr.22: 36930783-36930783 on Build GRCh38
1033900 c.916T>C p.C306R Chr.22: 36930734-36930734 on Build GRCh38
1033901 c.987G>T p.E329D Chr.22: 36930805-36930805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776142666 Chr.22: 36930771-36930771 on Build GRCh38
rs150251048 Chr.22: 36930759-36930759 on Build GRCh38
rs1021865587 Chr.22: 36930746-36930746 on Build GRCh38
rs144034620 Chr.22: 36930627-36930627 on Build GRCh38
rs116739204 Chr.22: 36930636-36930636 on Build GRCh38
rs781530019 Chr.22: 36930715-36930715 on Build GRCh38
rs576486802 Chr.22: 36930710-36930710 on Build GRCh38
rs201954835 Chr.22: 36930674-36930674 on Build GRCh38
rs189973117 Chr.22: 36930587-36930587 on Build GRCh38
rs758984857 Chr.22: 36930778-36930778 on Build GRCh38
rs201262017 Chr.22: 36930643-36930643 on Build GRCh38
rs151112655 Chr.22: 36930707-36930707 on Build GRCh38
rs571343472 Chr.22: 36930703-36930703 on Build GRCh38
rs745934088 Chr.22: 36930660-36930660 on Build GRCh38
rs373556286 Chr.22: 36930751-36930751 on Build GRCh38
rs759668933 Chr.22: 36930695-36930695 on Build GRCh38
rs928518258 Chr.22: 36930756-36930756 on Build GRCh38
rs760148608 Chr.22: 36930699-36930699 on Build GRCh38
rs370426702 Chr.22: 36930658-36930658 on Build GRCh38
rs761485907 Chr.22: 36930613-36930613 on Build GRCh38
rs768505622 Chr.22: 36930671-36930671 on Build GRCh38
rs757623965 Chr.22: 36930713-36930713 on Build GRCh38
rs778151079 Chr.22: 36930638-36930638 on Build GRCh38
rs761525371 Chr.22: 36930685-36930685 on Build GRCh38
rs117805308 Chr.22: 36930752-36930752 on Build GRCh38
rs746137268 Chr.22: 36930721-36930721 on Build GRCh38
rs539051663 Chr.22: 36930661-36930661 on Build GRCh38
rs764516861 Chr.22: 36930709-36930709 on Build GRCh38
rs267606239 Chr.22: 36930672-36930672 on Build GRCh38
rs749331765 Chr.22: 36930745-36930745 on Build GRCh38
rs767079000 Chr.22: 36930614-36930614 on Build GRCh38
rs368230425 Chr.22: 36930779-36930779 on Build GRCh38
rs138918081 Chr.22: 36930760-36930760 on Build GRCh38
rs375783135 Chr.22: 36930758-36930758 on Build GRCh38
rs1038250280 Chr.22: 36930612-36930612 on Build GRCh38
rs756424237 Chr.22: 36930727-36930727 on Build GRCh38
rs547780142 Chr.22: 36930609-36930609 on Build GRCh38
rs749198467 Chr.22: 36930664-36930664 on Build GRCh38
rs747969899 Chr.22: 36930757-36930757 on Build GRCh38
rs757436084 Chr.22: 36930654-36930654 on Build GRCh38
rs555140256 Chr.22: 36930766-36930766 on Build GRCh38
rs775542545 Chr.22: 36930662-36930662 on Build GRCh38
rs375358994 Chr.22: 36930628-36930628 on Build GRCh38
rs1030675779 Chr.22: 36930692-36930692 on Build GRCh38
rs984721671 Chr.22: 36930750-36930750 on Build GRCh38
rs765923544 Chr.22: 36930705-36930705 on Build GRCh38
rs1011820968 Chr.22: 36930588-36930588 on Build GRCh38
rs774127718 Chr.22: 36930677-36930677 on Build GRCh38
rs767143075 Chr.22: 36930687-36930687 on Build GRCh38

View Full Product Details