Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135546289-135546562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS6L
Entrez Gene ID
203522
Gene Name
integrator complex subunit 6 like
Gene Aliases
DDX26B
UniGene
Hs.496829
Gene Chromosome Location
Chr.X: 135520630-135582535 on Build GRCh38
Transcripts
NM_182540.4 XM_005262394.2 XM_006724740.2 XM_006724741.3 XM_011531311.2 XM_017029342.1 XM_017029343.1 XM_017029344.1 XM_017029345.1 XM_017029346.1 XM_017029347.1 XM_017029348.1 XM_017029349.1

SNP Detail

SNP ID Chromosome Location
rs782200014 Chr.X: 135546481-135546481 on Build GRCh38
rs375005472 Chr.X: 135546515-135546515 on Build GRCh38
rs782242788 Chr.X: 135546392-135546392 on Build GRCh38
rs782590261 Chr.X: 135546497-135546497 on Build GRCh38
rs782687977 Chr.X: 135546406-135546406 on Build GRCh38
rs782569118 Chr.X: 135546452-135546452 on Build GRCh38
rs782292702 Chr.X: 135546360-135546360 on Build GRCh38
rs1044325743 Chr.X: 135546382-135546382 on Build GRCh38
rs782000826 Chr.X: 135546373-135546373 on Build GRCh38
rs782408526 Chr.X: 135546364-135546364 on Build GRCh38
rs781950539 Chr.X: 135546389-135546389 on Build GRCh38
rs782745272 Chr.X: 135546377-135546377 on Build GRCh38
rs782182386 Chr.X: 135546505-135546505 on Build GRCh38
rs373950962 Chr.X: 135546376-135546376 on Build GRCh38
rs36018462 Chr.X: 135546495-135546496 on Build GRCh38
rs781847653 Chr.X: 135546490-135546490 on Build GRCh38
rs935719727 Chr.X: 135546477-135546477 on Build GRCh38
rs782192298 Chr.X: 135546480-135546481 on Build GRCh38
rs888759776 Chr.X: 135546523-135546523 on Build GRCh38
rs564780283 Chr.X: 135546480-135546480 on Build GRCh38
rs782480213 Chr.X: 135546492-135546492 on Build GRCh38
rs376860275 Chr.X: 135546393-135546393 on Build GRCh38
rs781924370 Chr.X: 135546339-135546339 on Build GRCh38
rs371250011 Chr.X: 135546410-135546410 on Build GRCh38

View Full Product Details