Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90044810-90045073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR1
Entrez Gene ID
1666
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
Gene Aliases
DECR, NADPH, SDR18C1
UniGene
Hs.492212
Gene Chromosome Location
Chr.8: 90001337-90052004 on Build GRCh38
Transcripts
NM_001359.1 XM_005250808.4 XM_011516880.2 XM_017013147.1 XM_017013148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6472731 c.781G>A p.E261K Chr.8: 90044891-90044891 on Build GRCh38
5875089 c.813delG p.T273fs*2 Chr.8: 90044923-90044923 on Build GRCh38
1178884 c.878A>G p.N293S Chr.8: 90044988-90044988 on Build GRCh38
5617471 c.811C>T p.L271L Chr.8: 90044921-90044921 on Build GRCh38
4509208 c.800C>T p.P267L Chr.8: 90044910-90044910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149423653 Chr.8: 90044854-90044854 on Build GRCh38
rs549904950 Chr.8: 90045002-90045002 on Build GRCh38
rs778599679 Chr.8: 90044942-90044942 on Build GRCh38
rs763068155 Chr.8: 90044868-90044868 on Build GRCh38
rs375878524 Chr.8: 90044923-90044923 on Build GRCh38
rs751827348 Chr.8: 90044891-90044891 on Build GRCh38
rs746916937 Chr.8: 90045020-90045021 on Build GRCh38
rs187196299 Chr.8: 90045001-90045001 on Build GRCh38
rs181636748 Chr.8: 90044973-90044973 on Build GRCh38
rs200547759 Chr.8: 90045035-90045035 on Build GRCh38
rs755964577 Chr.8: 90044919-90044919 on Build GRCh38
rs769018034 Chr.8: 90044968-90044968 on Build GRCh38
rs147668665 Chr.8: 90045003-90045003 on Build GRCh38
rs774728429 Chr.8: 90044996-90044996 on Build GRCh38
rs770805705 Chr.8: 90045021-90045021 on Build GRCh38
rs748663121 Chr.8: 90044836-90044836 on Build GRCh38
rs961907442 Chr.8: 90044957-90044957 on Build GRCh38
rs10235 Chr.8: 90044921-90044921 on Build GRCh38
rs747862157 Chr.8: 90044894-90044896 on Build GRCh38
rs762163418 Chr.8: 90044861-90044861 on Build GRCh38
rs201405525 Chr.8: 90044993-90044993 on Build GRCh38
rs1053370862 Chr.8: 90045013-90045013 on Build GRCh38
rs867052303 Chr.8: 90044909-90044909 on Build GRCh38
rs773740195 Chr.8: 90044987-90044987 on Build GRCh38
rs767362254 Chr.8: 90044913-90044913 on Build GRCh38
rs148549954 Chr.8: 90044970-90044970 on Build GRCh38
rs370430849 Chr.8: 90044961-90044961 on Build GRCh38
rs776269139 Chr.8: 90044941-90044941 on Build GRCh38
rs759764213 Chr.8: 90045000-90045000 on Build GRCh38
rs532651944 Chr.8: 90045028-90045028 on Build GRCh38
rs372913734 Chr.8: 90044918-90044918 on Build GRCh38
rs561353886 Chr.8: 90044939-90044939 on Build GRCh38
rs762073849 Chr.8: 90044985-90044985 on Build GRCh38
rs752383256 Chr.8: 90045022-90045022 on Build GRCh38
rs992422968 Chr.8: 90044951-90044951 on Build GRCh38
rs948412780 Chr.8: 90044976-90044976 on Build GRCh38
rs199693288 Chr.8: 90044968-90044969 on Build GRCh38
rs756813637 Chr.8: 90045045-90045045 on Build GRCh38
rs144788496 Chr.8: 90044862-90044862 on Build GRCh38
rs201871108 Chr.8: 90044837-90044837 on Build GRCh38
rs760492974 Chr.8: 90045026-90045026 on Build GRCh38
rs759462502 Chr.8: 90044901-90044901 on Build GRCh38
rs771832515 Chr.8: 90044929-90044929 on Build GRCh38
rs892523731 Chr.8: 90045019-90045019 on Build GRCh38
rs925902750 Chr.8: 90044999-90044999 on Build GRCh38
rs901163079 Chr.8: 90045038-90045038 on Build GRCh38
rs766749750 Chr.8: 90044873-90044873 on Build GRCh38

View Full Product Details