Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 22451154-22451381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA1
Entrez Gene ID
63951
Gene Name
DMRT like family A1
Gene Aliases
DMO, DMRT4
UniGene
Hs.371976
Gene Chromosome Location
Chr.9: 22446841-22452473 on Build GRCh38
Transcripts
NM_022160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3092369 c.793T>C p.S265P Chr.9: 22451189-22451189 on Build GRCh38
5803712 c.794C>T p.S265L Chr.9: 22451190-22451190 on Build GRCh38
6645705 c.960G>A p.P320P Chr.9: 22451356-22451356 on Build GRCh38
6344021 c.834G>T p.L278L Chr.9: 22451230-22451230 on Build GRCh38
4425581 c.794C>G p.S265* Chr.9: 22451190-22451190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936560925 Chr.9: 22451215-22451215 on Build GRCh38
rs745339156 Chr.9: 22451294-22451294 on Build GRCh38
rs202197256 Chr.9: 22451350-22451350 on Build GRCh38
rs758212329 Chr.9: 22451239-22451239 on Build GRCh38
rs777522680 Chr.9: 22451191-22451191 on Build GRCh38
rs752706554 Chr.9: 22451237-22451237 on Build GRCh38
rs780948456 Chr.9: 22451287-22451287 on Build GRCh38
rs142995523 Chr.9: 22451270-22451270 on Build GRCh38
rs756763306 Chr.9: 22451195-22451195 on Build GRCh38
rs368798749 Chr.9: 22451328-22451328 on Build GRCh38
rs201374002 Chr.9: 22451232-22451232 on Build GRCh38
rs1029635429 Chr.9: 22451212-22451212 on Build GRCh38
rs777036865 Chr.9: 22451211-22451211 on Build GRCh38
rs371171820 Chr.9: 22451344-22451344 on Build GRCh38
rs754863697 Chr.9: 22451183-22451183 on Build GRCh38
rs763988468 Chr.9: 22451256-22451256 on Build GRCh38
rs267602201 Chr.9: 22451304-22451304 on Build GRCh38
rs1006426815 Chr.9: 22451266-22451266 on Build GRCh38
rs137914267 Chr.9: 22451356-22451356 on Build GRCh38
rs962865767 Chr.9: 22451233-22451233 on Build GRCh38
rs771947601 Chr.9: 22451268-22451268 on Build GRCh38
rs61729546 Chr.9: 22451340-22451340 on Build GRCh38
rs1015081364 Chr.9: 22451267-22451267 on Build GRCh38
rs74769337 Chr.9: 22451228-22451228 on Build GRCh38
rs952326505 Chr.9: 22451259-22451259 on Build GRCh38
rs746675446 Chr.9: 22451193-22451193 on Build GRCh38
rs145152745 Chr.9: 22451278-22451278 on Build GRCh38
rs181512603 Chr.9: 22451185-22451185 on Build GRCh38
rs748471195 Chr.9: 22451220-22451220 on Build GRCh38
rs949805564 Chr.9: 22451354-22451354 on Build GRCh38
rs772447910 Chr.9: 22451222-22451222 on Build GRCh38
rs776947180 Chr.9: 22451349-22451349 on Build GRCh38
rs1001508505 Chr.9: 22451244-22451244 on Build GRCh38
rs771032631 Chr.9: 22451348-22451348 on Build GRCh38
rs369778210 Chr.9: 22451358-22451358 on Build GRCh38
rs777892111 Chr.9: 22451281-22451283 on Build GRCh38
rs760888914 Chr.9: 22451227-22451227 on Build GRCh38
rs34324424 Chr.9: 22451230-22451230 on Build GRCh38
rs758459142 Chr.9: 22451178-22451179 on Build GRCh38
rs139030144 Chr.9: 22451338-22451338 on Build GRCh38
rs368428828 Chr.9: 22451217-22451217 on Build GRCh38
rs1033333947 Chr.9: 22451188-22451188 on Build GRCh38
rs1034283321 Chr.9: 22451248-22451248 on Build GRCh38
rs1041549610 Chr.9: 22451182-22451182 on Build GRCh38
rs150709609 Chr.9: 22451343-22451343 on Build GRCh38
rs745441990 Chr.9: 22451204-22451204 on Build GRCh38
rs773362908 Chr.9: 22451223-22451223 on Build GRCh38
rs779238351 Chr.9: 22451324-22451324 on Build GRCh38
rs148944998 Chr.9: 22451186-22451186 on Build GRCh38
rs755681561 Chr.9: 22451301-22451301 on Build GRCh38
rs143713234 Chr.9: 22451203-22451203 on Build GRCh38
rs753740634 Chr.9: 22451181-22451181 on Build GRCh38

View Full Product Details