Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 88114523-88114784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
UniGene
Hs.256398
Gene Chromosome Location
Chr.7: 87934143-88202889 on Build GRCh38
Transcripts
NM_001324417.1 NM_001324418.1 NM_001324419.1 NM_001324420.1 NM_001324421.1 NM_004194.4 NM_016351.5 NM_021721.4 NM_021722.5 NM_021723.4 XM_005250445.3 XM_006716028.3 XM_006716029.3 XM_011516318.1 XM_011516319.2 XM_011516320.2 XM_011516321.2 XM_011516322.1 XM_011516323.1 XM_011516324.1 XM_017012329.1 XM_017012330.1 XM_017012331.1 XM_017012332.1 XM_017012333.1 XM_017012334.1 XM_017012335.1 XM_017012336.1 XM_017012337.1 XM_017012338.1 XM_017012339.1 XM_017012341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778646 c.507C>G p.L169L Chr.7: 88114617-88114617 on Build GRCh38
1181800 c.478A>G p.M160V Chr.7: 88114588-88114588 on Build GRCh38
1092687 c.525T>C p.N175N Chr.7: 88114635-88114635 on Build GRCh38
1181799 c.478A>G p.M160V Chr.7: 88114588-88114588 on Build GRCh38
1092686 c.525T>C p.N175N Chr.7: 88114635-88114635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546677539 Chr.7: 88114719-88114720 on Build GRCh38
rs754955153 Chr.7: 88114687-88114687 on Build GRCh38
rs753887308 Chr.7: 88114644-88114644 on Build GRCh38
rs181849057 Chr.7: 88114742-88114742 on Build GRCh38
rs576418684 Chr.7: 88114731-88114731 on Build GRCh38
rs761915231 Chr.7: 88114645-88114645 on Build GRCh38
rs775819084 Chr.7: 88114619-88114619 on Build GRCh38
rs753946917 Chr.7: 88114563-88114563 on Build GRCh38
rs764216731 Chr.7: 88114559-88114559 on Build GRCh38
rs781203785 Chr.7: 88114689-88114689 on Build GRCh38
rs774878910 Chr.7: 88114604-88114604 on Build GRCh38
rs1040797874 Chr.7: 88114626-88114626 on Build GRCh38
rs146241988 Chr.7: 88114575-88114575 on Build GRCh38
rs1029157562 Chr.7: 88114699-88114700 on Build GRCh38
rs751567619 Chr.7: 88114666-88114666 on Build GRCh38
rs769570093 Chr.7: 88114595-88114595 on Build GRCh38
rs371984804 Chr.7: 88114600-88114600 on Build GRCh38
rs368408489 Chr.7: 88114599-88114599 on Build GRCh38
rs371164835 Chr.7: 88114547-88114547 on Build GRCh38
rs781115188 Chr.7: 88114580-88114580 on Build GRCh38
rs371816665 Chr.7: 88114576-88114576 on Build GRCh38
rs796710750 Chr.7: 88114721-88114722 on Build GRCh38
rs34182051 Chr.7: 88114722-88114722 on Build GRCh38
rs772472226 Chr.7: 88114614-88114614 on Build GRCh38
rs752954692 Chr.7: 88114548-88114548 on Build GRCh38
rs931227880 Chr.7: 88114571-88114571 on Build GRCh38
rs779993008 Chr.7: 88114567-88114567 on Build GRCh38
rs199829685 Chr.7: 88114577-88114577 on Build GRCh38
rs138682284 Chr.7: 88114720-88114721 on Build GRCh38
rs764243604 Chr.7: 88114638-88114638 on Build GRCh38
rs34003748 Chr.7: 88114721-88114722 on Build GRCh38
rs752476900 Chr.7: 88114693-88114693 on Build GRCh38
rs760954016 Chr.7: 88114632-88114632 on Build GRCh38
rs374662169 Chr.7: 88114589-88114589 on Build GRCh38
rs765201623 Chr.7: 88114664-88114664 on Build GRCh38
rs759925724 Chr.7: 88114606-88114606 on Build GRCh38
rs900936099 Chr.7: 88114713-88114713 on Build GRCh38
rs373691038 Chr.7: 88114558-88114558 on Build GRCh38

View Full Product Details