Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44773047-44773296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
UniGene
Hs.6088
Gene Chromosome Location
Chr.17: 44759037-44781846 on Build GRCh38
Transcripts
NM_001318933.1 NM_002390.5 XM_005257373.3 XM_017024647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3795678 c.843C>G p.L281L Chr.17: 44773278-44773278 on Build GRCh38
3191455 c.820G>A p.D274N Chr.17: 44773080-44773080 on Build GRCh38
1710377 c.803C>T p.S268F Chr.17: 44773063-44773063 on Build GRCh38
3518129 c.835G>A p.E279K Chr.17: 44773270-44773270 on Build GRCh38
6029523 c.825+5G>A p.? Chr.17: 44773090-44773090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886417733 Chr.17: 44773091-44773091 on Build GRCh38
rs370959628 Chr.17: 44773250-44773250 on Build GRCh38
rs188421925 Chr.17: 44773237-44773237 on Build GRCh38
rs1004842676 Chr.17: 44773096-44773096 on Build GRCh38
rs766085576 Chr.17: 44773223-44773223 on Build GRCh38
rs533350309 Chr.17: 44773198-44773198 on Build GRCh38
rs749047003 Chr.17: 44773117-44773117 on Build GRCh38
rs765551742 Chr.17: 44773124-44773125 on Build GRCh38
rs769956251 Chr.17: 44773104-44773104 on Build GRCh38
rs1034619399 Chr.17: 44773199-44773199 on Build GRCh38
rs773047823 Chr.17: 44773133-44773133 on Build GRCh38
rs775179932 Chr.17: 44773105-44773105 on Build GRCh38
rs750094659 Chr.17: 44773073-44773073 on Build GRCh38
rs753006439 Chr.17: 44773234-44773234 on Build GRCh38
rs900583261 Chr.17: 44773066-44773066 on Build GRCh38
rs757327904 Chr.17: 44773095-44773095 on Build GRCh38
rs73984069 Chr.17: 44773165-44773165 on Build GRCh38
rs140421831 Chr.17: 44773079-44773079 on Build GRCh38
rs767104587 Chr.17: 44773127-44773127 on Build GRCh38
rs760976297 Chr.17: 44773091-44773092 on Build GRCh38
rs781437125 Chr.17: 44773097-44773097 on Build GRCh38
rs1037670854 Chr.17: 44773182-44773182 on Build GRCh38
rs377086702 Chr.17: 44773123-44773123 on Build GRCh38
rs764394340 Chr.17: 44773095-44773096 on Build GRCh38
rs746072579 Chr.17: 44773101-44773101 on Build GRCh38
rs1001371761 Chr.17: 44773196-44773196 on Build GRCh38
rs747118579 Chr.17: 44773082-44773082 on Build GRCh38
rs942026332 Chr.17: 44773179-44773179 on Build GRCh38
rs760421937 Chr.17: 44773215-44773215 on Build GRCh38
rs756333782 Chr.17: 44773275-44773275 on Build GRCh38
rs1057337412 Chr.17: 44773221-44773221 on Build GRCh38
rs750464001 Chr.17: 44773241-44773241 on Build GRCh38
rs745908481 Chr.17: 44773251-44773251 on Build GRCh38
rs757431453 Chr.17: 44773125-44773125 on Build GRCh38
rs543544885 Chr.17: 44773235-44773235 on Build GRCh38
rs201078446 Chr.17: 44773125-44773125 on Build GRCh38
rs1001596303 Chr.17: 44773206-44773206 on Build GRCh38
rs764578305 Chr.17: 44773236-44773236 on Build GRCh38
rs184207190 Chr.17: 44773080-44773080 on Build GRCh38
rs757381119 Chr.17: 44773241-44773241 on Build GRCh38
rs754022918 Chr.17: 44773107-44773110 on Build GRCh38
rs907147763 Chr.17: 44773170-44773170 on Build GRCh38
rs895967044 Chr.17: 44773243-44773243 on Build GRCh38
rs1010520122 Chr.17: 44773272-44773272 on Build GRCh38

View Full Product Details