Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207469043-207469316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4764489 c.781C>T p.Q261* Chr.1: 207469196-207469196 on Build GRCh38
5039782 c.784G>A p.G262R Chr.1: 207469199-207469199 on Build GRCh38
5845303 c.754C>T p.P252S Chr.1: 207469169-207469169 on Build GRCh38
4027604 c.788T>C p.V263A Chr.1: 207469203-207469203 on Build GRCh38
7214941 c.735-8G>A p.? Chr.1: 207469142-207469142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778859257 Chr.1: 207469251-207469251 on Build GRCh38
rs754918026 Chr.1: 207469122-207469122 on Build GRCh38
rs745918801 Chr.1: 207469150-207469150 on Build GRCh38
rs1030733127 Chr.1: 207469199-207469199 on Build GRCh38
rs763512398 Chr.1: 207469166-207469166 on Build GRCh38
rs781540705 Chr.1: 207469145-207469145 on Build GRCh38
rs769307763 Chr.1: 207469167-207469167 on Build GRCh38
rs771427606 Chr.1: 207469281-207469282 on Build GRCh38
rs759180891 Chr.1: 207469182-207469182 on Build GRCh38
rs200947637 Chr.1: 207469120-207469120 on Build GRCh38
rs80315759 Chr.1: 207469081-207469081 on Build GRCh38
rs750148498 Chr.1: 207469202-207469202 on Build GRCh38
rs113013210 Chr.1: 207469248-207469248 on Build GRCh38
rs150622293 Chr.1: 207469155-207469155 on Build GRCh38
rs989140673 Chr.1: 207469279-207469279 on Build GRCh38
rs771024465 Chr.1: 207469142-207469142 on Build GRCh38
rs1028968402 Chr.1: 207469077-207469077 on Build GRCh38
rs369546456 Chr.1: 207469152-207469152 on Build GRCh38
rs747982668 Chr.1: 207469132-207469132 on Build GRCh38
rs200630015 Chr.1: 207469123-207469123 on Build GRCh38
rs752482252 Chr.1: 207469273-207469273 on Build GRCh38
rs779846456 Chr.1: 207469119-207469119 on Build GRCh38
rs765852435 Chr.1: 207469204-207469204 on Build GRCh38
rs774836892 Chr.1: 207469175-207469175 on Build GRCh38
rs749871372 Chr.1: 207469106-207469106 on Build GRCh38
rs767250244 Chr.1: 207469100-207469100 on Build GRCh38
rs546816560 Chr.1: 207469115-207469115 on Build GRCh38
rs61754519 Chr.1: 207469178-207469178 on Build GRCh38
rs767073038 Chr.1: 207469197-207469197 on Build GRCh38
rs753380303 Chr.1: 207469224-207469224 on Build GRCh38

View Full Product Details