Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1419349-1419574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD65
Entrez Gene ID
441869
Gene Name
ankyrin repeat domain 65
Gene Aliases
-
UniGene
Hs.381222
Gene Chromosome Location
Chr.1: 1418420-1422561 on Build GRCh38
Transcripts
NM_001145210.2 NM_001243535.1 NM_001243536.1 XM_005244752.4 XM_006710647.3 XM_011541489.2 XM_017001324.1 XM_017001325.1 XM_017001326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305933 c.789C>A p.G263G Chr.1: 1419511-1419511 on Build GRCh38
6252982 c.919C>A p.H307N Chr.1: 1419381-1419381 on Build GRCh38
3688953 c.890C>T p.A297V Chr.1: 1419410-1419410 on Build GRCh38
4558677 c.776G>A p.G259E Chr.1: 1419524-1419524 on Build GRCh38
4951506 c.882G>C p.E294D Chr.1: 1419418-1419418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544484967 Chr.1: 1419436-1419436 on Build GRCh38
rs866962764 Chr.1: 1419474-1419474 on Build GRCh38
rs755685561 Chr.1: 1419384-1419384 on Build GRCh38
rs779213980 Chr.1: 1419410-1419410 on Build GRCh38
rs749112189 Chr.1: 1419465-1419465 on Build GRCh38
rs149947983 Chr.1: 1419501-1419501 on Build GRCh38
rs1044780746 Chr.1: 1419409-1419409 on Build GRCh38
rs552630536 Chr.1: 1419376-1419376 on Build GRCh38
rs755585269 Chr.1: 1419388-1419388 on Build GRCh38
rs780567034 Chr.1: 1419393-1419393 on Build GRCh38
rs939570700 Chr.1: 1419525-1419525 on Build GRCh38
rs921825562 Chr.1: 1419368-1419368 on Build GRCh38
rs989737959 Chr.1: 1419524-1419524 on Build GRCh38
rs181085315 Chr.1: 1419382-1419382 on Build GRCh38
rs755376821 Chr.1: 1419407-1419407 on Build GRCh38
rs140747215 Chr.1: 1419464-1419464 on Build GRCh38
rs760197934 Chr.1: 1419504-1419504 on Build GRCh38
rs886203633 Chr.1: 1419400-1419400 on Build GRCh38
rs879372192 Chr.1: 1419508-1419508 on Build GRCh38
rs989522604 Chr.1: 1419485-1419485 on Build GRCh38
rs1003835124 Chr.1: 1419426-1419426 on Build GRCh38
rs772120392 Chr.1: 1419448-1419448 on Build GRCh38
rs893962628 Chr.1: 1419375-1419375 on Build GRCh38
rs185980964 Chr.1: 1419420-1419420 on Build GRCh38
rs544720725 Chr.1: 1419399-1419399 on Build GRCh38
rs560402922 Chr.1: 1419515-1419515 on Build GRCh38
rs867172596 Chr.1: 1419381-1419381 on Build GRCh38
rs574552814 Chr.1: 1419468-1419468 on Build GRCh38
rs767193977 Chr.1: 1419491-1419491 on Build GRCh38
rs954747785 Chr.1: 1419463-1419463 on Build GRCh38
rs753387302 Chr.1: 1419533-1419533 on Build GRCh38
rs963203424 Chr.1: 1419421-1419421 on Build GRCh38
rs947819194 Chr.1: 1419543-1419543 on Build GRCh38
rs145112172 Chr.1: 1419540-1419540 on Build GRCh38
rs773175407 Chr.1: 1419461-1419461 on Build GRCh38
rs906765623 Chr.1: 1419415-1419415 on Build GRCh38
rs761573390 Chr.1: 1419480-1419480 on Build GRCh38
rs1016129563 Chr.1: 1419408-1419408 on Build GRCh38
rs772807396 Chr.1: 1419497-1419497 on Build GRCh38
rs774071232 Chr.1: 1419469-1419469 on Build GRCh38
rs377079302 Chr.1: 1419514-1419514 on Build GRCh38
rs751226837 Chr.1: 1419369-1419369 on Build GRCh38
rs906746834 Chr.1: 1419390-1419390 on Build GRCh38
rs867093491 Chr.1: 1419526-1419526 on Build GRCh38
rs575134240 Chr.1: 1419428-1419428 on Build GRCh38
rs749981272 Chr.1: 1419387-1419387 on Build GRCh38
rs1002547176 Chr.1: 1419397-1419397 on Build GRCh38

View Full Product Details