Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 105472515-105472766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAOA
Entrez Gene ID
267012
Gene Name
D-amino acid oxidase activator
Gene Aliases
LG72, SG72
UniGene
Hs.381382
Gene Chromosome Location
Chr.13: 105465867-105491034 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

Gene Symbol
DAOA-AS1
Entrez Gene ID
282706
Gene Name
DAOA antisense RNA 1
Gene Aliases
DAOA-AS, DAOAAS, G30
UniGene
Hs.558713
Gene Chromosome Location
Chr.13: 105459055-105505681 on Build GRCh38
Transcripts
NM_001161812.1 NM_001161814.1 NM_172370.4 XM_005254042.1 NR_040247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5036366 c.161C>T p.T54M Chr.13: 105472565-105472565 on Build GRCh38
3793097 c.222G>C p.Q74H Chr.13: 105472626-105472626 on Build GRCh38
6205511 c.134-10A>G p.? Chr.13: 105472528-105472528 on Build GRCh38
4045803 c.151G>T p.G51* Chr.13: 105472555-105472555 on Build GRCh38
5816256 c.278C>A p.A93E Chr.13: 105472682-105472682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755397146 Chr.13: 105472627-105472627 on Build GRCh38
rs748764075 Chr.13: 105472567-105472567 on Build GRCh38
rs140575409 Chr.13: 105472582-105472582 on Build GRCh38
rs776469368 Chr.13: 105472649-105472649 on Build GRCh38
rs201206506 Chr.13: 105472602-105472602 on Build GRCh38
rs200207534 Chr.13: 105472648-105472648 on Build GRCh38
rs916496022 Chr.13: 105472728-105472728 on Build GRCh38
rs761912948 Chr.13: 105472549-105472549 on Build GRCh38
rs780275757 Chr.13: 105472561-105472561 on Build GRCh38
rs751993855 Chr.13: 105472565-105472565 on Build GRCh38
rs768067587 Chr.13: 105472631-105472631 on Build GRCh38
rs72549478 Chr.13: 105472644-105472644 on Build GRCh38
rs1011528713 Chr.13: 105472643-105472643 on Build GRCh38
rs745423764 Chr.13: 105472641-105472641 on Build GRCh38
rs200951630 Chr.13: 105472606-105472606 on Build GRCh38
rs150372458 Chr.13: 105472736-105472736 on Build GRCh38
rs753155050 Chr.13: 105472710-105472710 on Build GRCh38
rs751934884 Chr.13: 105472706-105472707 on Build GRCh38
rs529535520 Chr.13: 105472711-105472711 on Build GRCh38
rs758675229 Chr.13: 105472555-105472555 on Build GRCh38
rs767978071 Chr.13: 105472709-105472709 on Build GRCh38
rs890016634 Chr.13: 105472646-105472646 on Build GRCh38
rs371012913 Chr.13: 105472651-105472651 on Build GRCh38
rs762609736 Chr.13: 105472685-105472685 on Build GRCh38
rs753267594 Chr.13: 105472632-105472632 on Build GRCh38
rs778537778 Chr.13: 105472640-105472640 on Build GRCh38
rs746847560 Chr.13: 105472598-105472598 on Build GRCh38
rs770567433 Chr.13: 105472574-105472574 on Build GRCh38
rs766677382 Chr.13: 105472689-105472690 on Build GRCh38
rs763053023 Chr.13: 105472612-105472612 on Build GRCh38
rs9558562 Chr.13: 105472588-105472588 on Build GRCh38
rs769762539 Chr.13: 105472663-105472663 on Build GRCh38
rs187721661 Chr.13: 105472596-105472596 on Build GRCh38
rs115136412 Chr.13: 105472692-105472692 on Build GRCh38
rs72549477 Chr.13: 105472566-105472566 on Build GRCh38
rs751822351 Chr.13: 105472620-105472620 on Build GRCh38
rs773346214 Chr.13: 105472610-105472610 on Build GRCh38
rs999559381 Chr.13: 105472686-105472686 on Build GRCh38
rs766671928 Chr.13: 105472615-105472615 on Build GRCh38
rs968143131 Chr.13: 105472666-105472666 on Build GRCh38
rs1022422500 Chr.13: 105472659-105472659 on Build GRCh38
rs763116465 Chr.13: 105472664-105472664 on Build GRCh38
rs201139062 Chr.13: 105472605-105472605 on Build GRCh38
rs765289215 Chr.13: 105472608-105472608 on Build GRCh38
rs72549479 Chr.13: 105472693-105472693 on Build GRCh38
rs901500344 Chr.13: 105472619-105472619 on Build GRCh38
rs367543079 Chr.13: 105472657-105472657 on Build GRCh38
rs72549480 Chr.13: 105472739-105472739 on Build GRCh38
rs367543080 Chr.13: 105472675-105472675 on Build GRCh38
rs745513380 Chr.13: 105472585-105472585 on Build GRCh38
rs992098530 Chr.13: 105472716-105472716 on Build GRCh38
rs753866109 Chr.13: 105472545-105472545 on Build GRCh38
rs765378685 Chr.13: 105472550-105472550 on Build GRCh38
rs772744576 Chr.13: 105472643-105472644 on Build GRCh38
rs756755022 Chr.13: 105472636-105472636 on Build GRCh38
rs750659467 Chr.13: 105472552-105472552 on Build GRCh38
rs764720402 Chr.13: 105472714-105472714 on Build GRCh38

View Full Product Details