Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62517384-62517609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHNAK
Entrez Gene ID
79026
Gene Name
AHNAK nucleoprotein
Gene Aliases
AHNAKRS
UniGene
Hs.502756
Gene Chromosome Location
Chr.11: 62433542-62546860 on Build GRCh38
Transcripts
NM_001620.2 NM_024060.3 XM_005274240.1 XM_005274241.2 XM_005274245.1 XM_017018270.1 XM_017018271.1 XM_017018272.1 XM_017018273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
689241 c.16811C>T p.S5604F Chr.11: 62517606-62517606 on Build GRCh38
6596903 c.16857C>A p.F5619L Chr.11: 62517560-62517560 on Build GRCh38
1355332 c.16969A>C p.T5657P Chr.11: 62517448-62517448 on Build GRCh38
7127081 c.16992C>A p.P5664P Chr.11: 62517425-62517425 on Build GRCh38
4842403 c.16920G>A p.L5640L Chr.11: 62517497-62517497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970551403 Chr.11: 62517458-62517458 on Build GRCh38
rs774860860 Chr.11: 62517525-62517525 on Build GRCh38
rs748428563 Chr.11: 62517489-62517489 on Build GRCh38
rs773733385 Chr.11: 62517518-62517518 on Build GRCh38
rs780039010 Chr.11: 62517477-62517477 on Build GRCh38
rs1048415188 Chr.11: 62517588-62517588 on Build GRCh38
rs751300626 Chr.11: 62517435-62517435 on Build GRCh38
rs763873648 Chr.11: 62517428-62517428 on Build GRCh38
rs566869163 Chr.11: 62517426-62517426 on Build GRCh38
rs757127540 Chr.11: 62517437-62517437 on Build GRCh38
rs753836595 Chr.11: 62517583-62517583 on Build GRCh38
rs764789867 Chr.11: 62517417-62517417 on Build GRCh38
rs768743408 Chr.11: 62517479-62517479 on Build GRCh38
rs762583341 Chr.11: 62517526-62517526 on Build GRCh38
rs779241308 Chr.11: 62517480-62517480 on Build GRCh38
rs761407110 Chr.11: 62517546-62517546 on Build GRCh38
rs540112394 Chr.11: 62517405-62517405 on Build GRCh38
rs371825003 Chr.11: 62517406-62517406 on Build GRCh38
rs755295460 Chr.11: 62517537-62517537 on Build GRCh38
rs750220866 Chr.11: 62517572-62517572 on Build GRCh38
rs139145890 Chr.11: 62517443-62517443 on Build GRCh38
rs749678925 Chr.11: 62517478-62517478 on Build GRCh38
rs767255176 Chr.11: 62517557-62517557 on Build GRCh38
rs143067494 Chr.11: 62517498-62517498 on Build GRCh38
rs11555628 Chr.11: 62517416-62517416 on Build GRCh38
rs756071881 Chr.11: 62517447-62517447 on Build GRCh38
rs761207153 Chr.11: 62517523-62517523 on Build GRCh38
rs375509429 Chr.11: 62517573-62517573 on Build GRCh38
rs751267774 Chr.11: 62517527-62517527 on Build GRCh38

View Full Product Details