Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16501415-16501688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599036 c.689G>C p.R230T Chr.19: 16501481-16501481 on Build GRCh38
6614882 c.723T>C p.A241A Chr.19: 16501515-16501515 on Build GRCh38
4623853 c.653T>C p.M218T Chr.19: 16501445-16501445 on Build GRCh38
6614883 c.718A>G p.S240G Chr.19: 16501510-16501510 on Build GRCh38
417720 c.663G>T p.L221F Chr.19: 16501455-16501455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008847775 Chr.19: 16501601-16501601 on Build GRCh38
rs938368853 Chr.19: 16501439-16501439 on Build GRCh38
rs139945315 Chr.19: 16501588-16501588 on Build GRCh38
rs748263116 Chr.19: 16501498-16501498 on Build GRCh38
rs760914620 Chr.19: 16501600-16501600 on Build GRCh38
rs964750256 Chr.19: 16501660-16501660 on Build GRCh38
rs756715466 Chr.19: 16501483-16501484 on Build GRCh38
rs756808963 Chr.19: 16501533-16501533 on Build GRCh38
rs778526412 Chr.19: 16501585-16501585 on Build GRCh38
rs765452090 Chr.19: 16501528-16501528 on Build GRCh38
rs745434284 Chr.19: 16501658-16501658 on Build GRCh38
rs772218577 Chr.19: 16501512-16501512 on Build GRCh38
rs529187397 Chr.19: 16501532-16501532 on Build GRCh38
rs751027426 Chr.19: 16501484-16501485 on Build GRCh38
rs749152333 Chr.19: 16501480-16501480 on Build GRCh38
rs755660084 Chr.19: 16501551-16501551 on Build GRCh38
rs767164001 Chr.19: 16501536-16501536 on Build GRCh38
rs372943238 Chr.19: 16501458-16501458 on Build GRCh38
rs780717311 Chr.19: 16501532-16501535 on Build GRCh38
rs761807578 Chr.19: 16501570-16501575 on Build GRCh38
rs7251978 Chr.19: 16501554-16501554 on Build GRCh38
rs749990577 Chr.19: 16501550-16501550 on Build GRCh38
rs777482076 Chr.19: 16501470-16501472 on Build GRCh38
rs373713416 Chr.19: 16501518-16501518 on Build GRCh38
rs756795441 Chr.19: 16501468-16501468 on Build GRCh38
rs569082927 Chr.19: 16501584-16501584 on Build GRCh38
rs745819965 Chr.19: 16501477-16501477 on Build GRCh38
rs201607227 Chr.19: 16501558-16501558 on Build GRCh38
rs768336435 Chr.19: 16501493-16501493 on Build GRCh38
rs989536958 Chr.19: 16501665-16501665 on Build GRCh38
rs777109146 Chr.19: 16501524-16501524 on Build GRCh38
rs35035783 Chr.19: 16501557-16501558 on Build GRCh38
rs772136839 Chr.19: 16501586-16501590 on Build GRCh38
rs192665443 Chr.19: 16501655-16501655 on Build GRCh38
rs753655864 Chr.19: 16501570-16501570 on Build GRCh38
rs774641690 Chr.19: 16501494-16501494 on Build GRCh38
rs745475290 Chr.19: 16501544-16501545 on Build GRCh38
rs754819433 Chr.19: 16501574-16501574 on Build GRCh38
rs200970238 Chr.19: 16501479-16501479 on Build GRCh38
rs773331330 Chr.19: 16501599-16501600 on Build GRCh38
rs188780108 Chr.19: 16501519-16501519 on Build GRCh38
rs754243003 Chr.19: 16501454-16501454 on Build GRCh38
rs796894266 Chr.19: 16501558-16501559 on Build GRCh38
rs756073383 Chr.19: 16501478-16501478 on Build GRCh38
rs571963647 Chr.19: 16501558-16501559 on Build GRCh38
rs866789895 Chr.19: 16501538-16501538 on Build GRCh38
rs763865304 Chr.19: 16501464-16501464 on Build GRCh38
rs201664638 Chr.19: 16501487-16501487 on Build GRCh38
rs751166122 Chr.19: 16501467-16501467 on Build GRCh38
rs367856596 Chr.19: 16501525-16501525 on Build GRCh38
rs751236926 Chr.19: 16501530-16501530 on Build GRCh38
rs773237666 Chr.19: 16501513-16501513 on Build GRCh38

View Full Product Details