Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19707041-19707268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KNOP1
Entrez Gene ID
400506
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
101F10.1, C16orf88, FAM191A, TSG118
UniGene
Hs.585209
Gene Chromosome Location
Chr.16: 19706347-19718240 on Build GRCh38
Transcripts
NM_001012991.2 XM_005255317.4 XM_006721046.3 XM_011545855.1 XM_017023231.1 XM_017023232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376318 c.1237C>T p.Q413* Chr.16: 19707050-19707050 on Build GRCh38
6697598 c.1180G>A p.A394T Chr.16: 19707107-19707107 on Build GRCh38
6697599 c.1223C>T p.A408V Chr.16: 19707064-19707064 on Build GRCh38
4667020 c.1097G>A p.G366D Chr.16: 19707190-19707190 on Build GRCh38
4667021 c.1095T>G p.A365A Chr.16: 19707192-19707192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372826065 Chr.16: 19707085-19707085 on Build GRCh38
rs557299469 Chr.16: 19707211-19707211 on Build GRCh38
rs371652855 Chr.16: 19707140-19707140 on Build GRCh38
rs200107473 Chr.16: 19707173-19707173 on Build GRCh38
rs752586146 Chr.16: 19707196-19707196 on Build GRCh38
rs755086987 Chr.16: 19707060-19707060 on Build GRCh38
rs750556184 Chr.16: 19707131-19707131 on Build GRCh38
rs777015287 Chr.16: 19707078-19707078 on Build GRCh38
rs780354698 Chr.16: 19707244-19707244 on Build GRCh38
rs546877998 Chr.16: 19707094-19707094 on Build GRCh38
rs773998129 Chr.16: 19707191-19707191 on Build GRCh38
rs769144454 Chr.16: 19707148-19707148 on Build GRCh38
rs747407192 Chr.16: 19707076-19707076 on Build GRCh38
rs770377126 Chr.16: 19707172-19707172 on Build GRCh38
rs781666118 Chr.16: 19707155-19707155 on Build GRCh38
rs775965613 Chr.16: 19707074-19707074 on Build GRCh38
rs764121998 Chr.16: 19707199-19707199 on Build GRCh38
rs369140660 Chr.16: 19707063-19707063 on Build GRCh38
rs1019662165 Chr.16: 19707228-19707228 on Build GRCh38
rs761392487 Chr.16: 19707100-19707100 on Build GRCh38
rs762475556 Chr.16: 19707179-19707179 on Build GRCh38
rs778836920 Chr.16: 19707127-19707127 on Build GRCh38
rs984751326 Chr.16: 19707105-19707105 on Build GRCh38
rs535661119 Chr.16: 19707121-19707121 on Build GRCh38
rs972269334 Chr.16: 19707241-19707241 on Build GRCh38
rs1020109558 Chr.16: 19707206-19707206 on Build GRCh38
rs1021962513 Chr.16: 19707190-19707190 on Build GRCh38
rs770515113 Chr.16: 19707180-19707180 on Build GRCh38
rs760497941 Chr.16: 19707197-19707197 on Build GRCh38
rs370433328 Chr.16: 19707215-19707215 on Build GRCh38
rs965830534 Chr.16: 19707240-19707240 on Build GRCh38
rs779686034 Chr.16: 19707083-19707092 on Build GRCh38
rs369471895 Chr.16: 19707150-19707150 on Build GRCh38
rs202156332 Chr.16: 19707141-19707141 on Build GRCh38
rs373516039 Chr.16: 19707161-19707161 on Build GRCh38
rs568591720 Chr.16: 19707168-19707168 on Build GRCh38
rs200258818 Chr.16: 19707107-19707107 on Build GRCh38
rs758564085 Chr.16: 19707234-19707234 on Build GRCh38
rs766992464 Chr.16: 19707115-19707115 on Build GRCh38
rs758185394 Chr.16: 19707060-19707061 on Build GRCh38
rs758652987 Chr.16: 19707138-19707138 on Build GRCh38
rs564785182 Chr.16: 19707077-19707077 on Build GRCh38
rs768078982 Chr.16: 19707145-19707145 on Build GRCh38
rs751309837 Chr.16: 19707108-19707108 on Build GRCh38
rs754350042 Chr.16: 19707177-19707177 on Build GRCh38
rs746554706 Chr.16: 19707108-19707108 on Build GRCh38
rs766605973 Chr.16: 19707097-19707097 on Build GRCh38
rs764645476 Chr.16: 19707090-19707090 on Build GRCh38
rs767203767 Chr.16: 19707194-19707194 on Build GRCh38
rs759253330 Chr.16: 19707193-19707193 on Build GRCh38
rs767423892 Chr.16: 19707119-19707119 on Build GRCh38
rs201786605 Chr.16: 19707113-19707113 on Build GRCh38
rs781223728 Chr.16: 19707062-19707062 on Build GRCh38
rs752573375 Chr.16: 19707120-19707120 on Build GRCh38
rs373038571 Chr.16: 19707064-19707064 on Build GRCh38

View Full Product Details