Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11377818-11378071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPOR
Entrez Gene ID
2057
Gene Name
erythropoietin receptor
Gene Aliases
EPO-R
UniGene
Hs.631624
Gene Chromosome Location
Chr.19: 11377205-11384342 on Build GRCh38
Transcripts
NM_000121.3 NR_033663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587730 c.1483C>T p.P495S Chr.19: 11378028-11378028 on Build GRCh38
7113991 c.1460A>G p.N487S Chr.19: 11378051-11378051 on Build GRCh38
2814354 c.1488C>T p.A496A Chr.19: 11378023-11378023 on Build GRCh38
1480555 c.1498C>A p.L500M Chr.19: 11378013-11378013 on Build GRCh38
6429357 c.1483C>A p.P495T Chr.19: 11378028-11378028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747661837 Chr.19: 11377947-11377947 on Build GRCh38
rs947599616 Chr.19: 11377978-11377978 on Build GRCh38
rs770519263 Chr.19: 11377960-11377960 on Build GRCh38
rs369251866 Chr.19: 11377876-11377876 on Build GRCh38
rs182850257 Chr.19: 11377956-11377956 on Build GRCh38
rs908204648 Chr.19: 11377971-11377971 on Build GRCh38
rs775746449 Chr.19: 11377966-11377966 on Build GRCh38
rs765416476 Chr.19: 11377990-11377990 on Build GRCh38
rs149193129 Chr.19: 11378023-11378023 on Build GRCh38
rs781154864 Chr.19: 11377946-11377946 on Build GRCh38
rs750673369 Chr.19: 11377907-11377907 on Build GRCh38
rs112348054 Chr.19: 11377979-11377979 on Build GRCh38
rs1039655747 Chr.19: 11378009-11378009 on Build GRCh38
rs764553795 Chr.19: 11377975-11377975 on Build GRCh38
rs779330970 Chr.19: 11377905-11377905 on Build GRCh38
rs142094773 Chr.19: 11378049-11378049 on Build GRCh38
rs535127351 Chr.19: 11377891-11377891 on Build GRCh38
rs1031687698 Chr.19: 11377855-11377855 on Build GRCh38
rs200372382 Chr.19: 11378006-11378006 on Build GRCh38
rs780010936 Chr.19: 11377924-11377924 on Build GRCh38
rs915694421 Chr.19: 11377993-11377993 on Build GRCh38
rs987568893 Chr.19: 11377885-11377885 on Build GRCh38
rs768508781 Chr.19: 11377943-11377943 on Build GRCh38
rs375280404 Chr.19: 11378021-11378021 on Build GRCh38
rs765467666 Chr.19: 11377955-11377955 on Build GRCh38
rs758630505 Chr.19: 11377917-11377917 on Build GRCh38
rs533243191 Chr.19: 11377953-11377953 on Build GRCh38
rs753901743 Chr.19: 11377902-11377902 on Build GRCh38
rs748120978 Chr.19: 11378029-11378029 on Build GRCh38
rs758697083 Chr.19: 11378005-11378005 on Build GRCh38
rs757333864 Chr.19: 11377903-11377903 on Build GRCh38
rs62638745 Chr.19: 11378051-11378051 on Build GRCh38
rs369228469 Chr.19: 11378010-11378010 on Build GRCh38
rs377657947 Chr.19: 11378041-11378041 on Build GRCh38
rs988547452 Chr.19: 11377959-11377959 on Build GRCh38
rs761106522 Chr.19: 11377881-11377881 on Build GRCh38
rs147359717 Chr.19: 11377878-11377878 on Build GRCh38
rs752862659 Chr.19: 11378004-11378005 on Build GRCh38
rs367583626 Chr.19: 11377955-11377955 on Build GRCh38
rs199784763 Chr.19: 11377952-11377952 on Build GRCh38
rs762055342 Chr.19: 11377986-11377986 on Build GRCh38
rs746771835 Chr.19: 11377931-11377931 on Build GRCh38
rs751352150 Chr.19: 11378022-11378022 on Build GRCh38
rs200997864 Chr.19: 11377949-11377949 on Build GRCh38
rs552777813 Chr.19: 11377869-11377869 on Build GRCh38
rs981122712 Chr.19: 11377893-11377893 on Build GRCh38
rs770608744 Chr.19: 11378044-11378044 on Build GRCh38
rs776340905 Chr.19: 11378038-11378038 on Build GRCh38
rs761066937 Chr.19: 11377967-11377967 on Build GRCh38

View Full Product Details