Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33566116-33566353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD18B
Entrez Gene ID
441459
Gene Name
ankyrin repeat domain 18B
Gene Aliases
bA255A11.3, bA255A11.5
UniGene
Hs.493710
Gene Chromosome Location
Chr.9: 33524403-33578755 on Build GRCh38
Transcripts
NM_001244752.1 XM_011517881.2 XM_011517883.2 XM_011517885.2 XM_017014728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7355575 c.2324A>G p.N775S Chr.9: 33566268-33566268 on Build GRCh38
3763892 c.2292C>T p.A764A Chr.9: 33566236-33566236 on Build GRCh38
6332065 c.2292C>G p.A764A Chr.9: 33566236-33566236 on Build GRCh38
1636165 c.2318_2319insT p.V774fs*8 Chr.9: 33566262-33566263 on Build GRCh38
6264500 c.2341G>C p.E781Q Chr.9: 33566285-33566285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142124052 Chr.9: 33566268-33566268 on Build GRCh38
rs902346608 Chr.9: 33566257-33566257 on Build GRCh38
rs749313111 Chr.9: 33566175-33566175 on Build GRCh38
rs373114045 Chr.9: 33566312-33566312 on Build GRCh38
rs939343592 Chr.9: 33566155-33566155 on Build GRCh38
rs778817242 Chr.9: 33566200-33566200 on Build GRCh38
rs538597138 Chr.9: 33566187-33566187 on Build GRCh38
rs995820178 Chr.9: 33566316-33566316 on Build GRCh38
rs537865672 Chr.9: 33566288-33566288 on Build GRCh38
rs554573425 Chr.9: 33566289-33566289 on Build GRCh38
rs770620595 Chr.9: 33566277-33566277 on Build GRCh38
rs574459570 Chr.9: 33566302-33566302 on Build GRCh38
rs764490442 Chr.9: 33566291-33566291 on Build GRCh38
rs41306100 Chr.9: 33566315-33566315 on Build GRCh38
rs11791519 Chr.9: 33566181-33566181 on Build GRCh38
rs762044641 Chr.9: 33566309-33566309 on Build GRCh38
rs1055073663 Chr.9: 33566199-33566199 on Build GRCh38
rs965812983 Chr.9: 33566313-33566313 on Build GRCh38
rs747847710 Chr.9: 33566233-33566233 on Build GRCh38
rs62559879 Chr.9: 33566236-33566236 on Build GRCh38
rs772929260 Chr.9: 33566237-33566237 on Build GRCh38

View Full Product Details