Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70087997-70088226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
UniGene
Hs.503074
Gene Chromosome Location
Chr.11: 70078302-70189545 on Build GRCh38
Transcripts
NM_018043.5 NR_030691.1 XM_006718602.2 XM_006718604.2 XM_006718605.2 XM_011545121.2 XM_011545123.2 XM_011545124.2 XM_011545125.2 XM_011545126.2 XM_011545127.2 XM_011545128.2 XM_011545129.2 XM_011545131.2 XM_017017956.1 XM_017017957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3452669 c.321C>T p.L107L Chr.11: 70088048-70088048 on Build GRCh38
5373450 c.409G>A p.E137K Chr.11: 70088052-70088052 on Build GRCh38
5458276 c.393C>T p.Y131Y Chr.11: 70088036-70088036 on Build GRCh38
5373452 c.325G>A p.E109K Chr.11: 70088052-70088052 on Build GRCh38
5458278 c.309C>T p.Y103Y Chr.11: 70088036-70088036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868840034 Chr.11: 70088037-70088037 on Build GRCh38
rs905391800 Chr.11: 70088110-70088132 on Build GRCh38
rs140065933 Chr.11: 70088119-70088120 on Build GRCh38
rs1008403045 Chr.11: 70088039-70088039 on Build GRCh38
rs1011712698 Chr.11: 70088125-70088125 on Build GRCh38
rs781298945 Chr.11: 70088084-70088084 on Build GRCh38
rs751122973 Chr.11: 70088112-70088112 on Build GRCh38
rs928423982 Chr.11: 70088172-70088172 on Build GRCh38
rs931564749 Chr.11: 70088112-70088112 on Build GRCh38
rs757934686 Chr.11: 70088074-70088074 on Build GRCh38
rs753396669 Chr.11: 70088048-70088048 on Build GRCh38
rs370908183 Chr.11: 70088023-70088023 on Build GRCh38
rs200695647 Chr.11: 70088036-70088036 on Build GRCh38
rs199813787 Chr.11: 70088079-70088079 on Build GRCh38
rs765941119 Chr.11: 70088090-70088090 on Build GRCh38
rs955897664 Chr.11: 70088051-70088051 on Build GRCh38
rs887294520 Chr.11: 70088147-70088147 on Build GRCh38
rs761236285 Chr.11: 70088029-70088029 on Build GRCh38
rs866726371 Chr.11: 70088123-70088123 on Build GRCh38
rs938588376 Chr.11: 70088185-70088185 on Build GRCh38
rs542259870 Chr.11: 70088078-70088078 on Build GRCh38
rs12796368 Chr.11: 70088118-70088118 on Build GRCh38
rs377496693 Chr.11: 70088057-70088057 on Build GRCh38
rs370422713 Chr.11: 70088064-70088064 on Build GRCh38
rs1023024860 Chr.11: 70088134-70088134 on Build GRCh38
rs565054101 Chr.11: 70088187-70088187 on Build GRCh38
rs745756595 Chr.11: 70088102-70088102 on Build GRCh38
rs12785876 Chr.11: 70088117-70088117 on Build GRCh38
rs970550017 Chr.11: 70088145-70088145 on Build GRCh38
rs1047202058 Chr.11: 70088146-70088146 on Build GRCh38
rs111840361 Chr.11: 70088152-70088152 on Build GRCh38
rs761870791 Chr.11: 70088022-70088022 on Build GRCh38
rs754539827 Chr.11: 70088056-70088056 on Build GRCh38
rs572841890 Chr.11: 70088146-70088146 on Build GRCh38
rs777772321 Chr.11: 70088075-70088075 on Build GRCh38

View Full Product Details