Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50723910-50724159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC11A
Entrez Gene ID
6320
Gene Name
C-type lectin domain family 11 member A
Gene Aliases
CLECSF3, LSLCL, P47, SCGF
UniGene
Hs.512680
Gene Chromosome Location
Chr.19: 50723348-50725724 on Build GRCh38
Transcripts
NM_002975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4517219 c.298_299CC>TT p.P100F Chr.19: 50724055-50724056 on Build GRCh38
5056864 c.322G>A p.V108I Chr.19: 50724079-50724079 on Build GRCh38
4487702 c.321C>T p.I107I Chr.19: 50724078-50724078 on Build GRCh38
319482 c.247G>A p.G83R Chr.19: 50724004-50724004 on Build GRCh38
7186282 c.303T>A p.S101S Chr.19: 50724060-50724060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779411078 Chr.19: 50724056-50724056 on Build GRCh38
rs768355917 Chr.19: 50723938-50723939 on Build GRCh38
rs745903366 Chr.19: 50723960-50723960 on Build GRCh38
rs375845376 Chr.19: 50724083-50724083 on Build GRCh38
rs771970804 Chr.19: 50724061-50724061 on Build GRCh38
rs141783525 Chr.19: 50724066-50724066 on Build GRCh38
rs766185930 Chr.19: 50724113-50724113 on Build GRCh38
rs139906595 Chr.19: 50724014-50724014 on Build GRCh38
rs748590355 Chr.19: 50723941-50723941 on Build GRCh38
rs761600337 Chr.19: 50724078-50724078 on Build GRCh38
rs778470033 Chr.19: 50723951-50723951 on Build GRCh38
rs766188693 Chr.19: 50724030-50724030 on Build GRCh38
rs751072504 Chr.19: 50724106-50724106 on Build GRCh38
rs774318485 Chr.19: 50723959-50723959 on Build GRCh38
rs779346643 Chr.19: 50724133-50724133 on Build GRCh38
rs754158536 Chr.19: 50724116-50724116 on Build GRCh38
rs757998254 Chr.19: 50723930-50723930 on Build GRCh38
rs998514470 Chr.19: 50724097-50724097 on Build GRCh38
rs760440562 Chr.19: 50724111-50724117 on Build GRCh38
rs757720001 Chr.19: 50724048-50724048 on Build GRCh38
rs747681407 Chr.19: 50724014-50724015 on Build GRCh38
rs200240589 Chr.19: 50724102-50724102 on Build GRCh38
rs750923831 Chr.19: 50724134-50724134 on Build GRCh38
rs757636946 Chr.19: 50724118-50724118 on Build GRCh38
rs369513922 Chr.19: 50724001-50724001 on Build GRCh38
rs772287072 Chr.19: 50723965-50723965 on Build GRCh38
rs758516738 Chr.19: 50724136-50724136 on Build GRCh38
rs762600692 Chr.19: 50724095-50724095 on Build GRCh38
rs928103139 Chr.19: 50723994-50723994 on Build GRCh38
rs202169240 Chr.19: 50724046-50724046 on Build GRCh38
rs756530261 Chr.19: 50723942-50723942 on Build GRCh38
rs1054276806 Chr.19: 50724059-50724059 on Build GRCh38
rs767141899 Chr.19: 50724114-50724114 on Build GRCh38
rs938173535 Chr.19: 50723995-50723995 on Build GRCh38
rs776301150 Chr.19: 50724075-50724075 on Build GRCh38
rs982179501 Chr.19: 50723967-50723967 on Build GRCh38
rs868795626 Chr.19: 50724076-50724076 on Build GRCh38
rs746002325 Chr.19: 50724062-50724062 on Build GRCh38
rs759282601 Chr.19: 50724113-50724113 on Build GRCh38
rs779785903 Chr.19: 50723940-50723940 on Build GRCh38
rs760525115 Chr.19: 50723983-50723983 on Build GRCh38
rs776374380 Chr.19: 50724118-50724118 on Build GRCh38
rs368263729 Chr.19: 50724012-50724013 on Build GRCh38
rs963258000 Chr.19: 50723953-50723953 on Build GRCh38
rs761689226 Chr.19: 50724005-50724005 on Build GRCh38
rs768828418 Chr.19: 50724070-50724070 on Build GRCh38
rs768420995 Chr.19: 50723984-50723984 on Build GRCh38
rs775686052 Chr.19: 50723969-50723969 on Build GRCh38
rs771427477 Chr.19: 50723958-50723958 on Build GRCh38
rs762544046 Chr.19: 50724029-50724029 on Build GRCh38
rs1042644476 Chr.19: 50724067-50724067 on Build GRCh38
rs749725650 Chr.19: 50723952-50723952 on Build GRCh38
rs751233976 Chr.19: 50724045-50724045 on Build GRCh38
rs2303688 Chr.19: 50724068-50724068 on Build GRCh38
rs774012963 Chr.19: 50723949-50723950 on Build GRCh38
rs773173758 Chr.19: 50724104-50724109 on Build GRCh38

View Full Product Details