Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67279808-67280059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK2
Entrez Gene ID
156
Gene Name
G protein-coupled receptor kinase 2
Gene Aliases
ADRBK1, BARK1, BETA-ARK1
UniGene
Hs.83636
Gene Chromosome Location
Chr.11: 67266420-67286558 on Build GRCh38
Transcripts
NM_001619.3 XM_011544773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451234 c.472C>T p.R158* Chr.11: 67279869-67279869 on Build GRCh38
4574709 c.478G>T p.D160Y Chr.11: 67279875-67279875 on Build GRCh38
7341075 c.463C>G p.Q155E Chr.11: 67279860-67279860 on Build GRCh38
5341699 c.451G>A p.E151K Chr.11: 67279848-67279848 on Build GRCh38
7215305 c.481G>A p.V161M Chr.11: 67279878-67279878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036963105 Chr.11: 67280028-67280028 on Build GRCh38
rs535763507 Chr.11: 67279960-67279960 on Build GRCh38
rs143344653 Chr.11: 67279877-67279877 on Build GRCh38
rs747199888 Chr.11: 67279951-67279951 on Build GRCh38
rs769763763 Chr.11: 67279919-67279928 on Build GRCh38
rs761569320 Chr.11: 67279876-67279876 on Build GRCh38
rs3730310 Chr.11: 67279929-67279929 on Build GRCh38
rs755590208 Chr.11: 67279926-67279926 on Build GRCh38
rs377671393 Chr.11: 67279875-67279875 on Build GRCh38
rs375408675 Chr.11: 67279914-67279914 on Build GRCh38
rs939664565 Chr.11: 67280020-67280020 on Build GRCh38
rs762912207 Chr.11: 67279870-67279870 on Build GRCh38
rs763973251 Chr.11: 67279874-67279874 on Build GRCh38
rs1047485224 Chr.11: 67279853-67279853 on Build GRCh38
rs371585088 Chr.11: 67279944-67279944 on Build GRCh38
rs368729302 Chr.11: 67279931-67279931 on Build GRCh38
rs370934447 Chr.11: 67279894-67279894 on Build GRCh38
rs909554071 Chr.11: 67279998-67279998 on Build GRCh38
rs192113785 Chr.11: 67279921-67279921 on Build GRCh38
rs778311170 Chr.11: 67280005-67280005 on Build GRCh38
rs61763962 Chr.11: 67279986-67279986 on Build GRCh38
rs926431516 Chr.11: 67279953-67279953 on Build GRCh38
rs554168506 Chr.11: 67279968-67279968 on Build GRCh38
rs771530727 Chr.11: 67279848-67279848 on Build GRCh38
rs760863502 Chr.11: 67279843-67279843 on Build GRCh38
rs990764987 Chr.11: 67280023-67280023 on Build GRCh38
rs568332978 Chr.11: 67279930-67279930 on Build GRCh38
rs758902035 Chr.11: 67279945-67279945 on Build GRCh38
rs1004032340 Chr.11: 67279985-67279985 on Build GRCh38
rs748500568 Chr.11: 67279839-67279839 on Build GRCh38
rs765748675 Chr.11: 67279940-67279940 on Build GRCh38
rs773298228 Chr.11: 67279841-67279841 on Build GRCh38
rs770926241 Chr.11: 67279845-67279845 on Build GRCh38
rs772378377 Chr.11: 67279840-67279840 on Build GRCh38
rs776703373 Chr.11: 67279850-67279850 on Build GRCh38

View Full Product Details