Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100091419-100091664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3883926 c.773G>A p.R258Q Chr.7: 100091450-100091450 on Build GRCh38
6584693 c.760C>G p.H254D Chr.7: 100091437-100091437 on Build GRCh38
5452049 c.775_776delGA p.E259fs*22 Chr.7: 100091452-100091453 on Build GRCh38
3833658 c.793C>T p.H265Y Chr.7: 100091470-100091470 on Build GRCh38
1093943 c.832G>T p.D278Y Chr.7: 100091509-100091509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779075759 Chr.7: 100091523-100091523 on Build GRCh38
rs1054592563 Chr.7: 100091601-100091601 on Build GRCh38
rs764936878 Chr.7: 100091462-100091462 on Build GRCh38
rs369196927 Chr.7: 100091570-100091570 on Build GRCh38
rs906097475 Chr.7: 100091581-100091581 on Build GRCh38
rs773052192 Chr.7: 100091481-100091481 on Build GRCh38
rs878927857 Chr.7: 100091446-100091446 on Build GRCh38
rs770326508 Chr.7: 100091559-100091559 on Build GRCh38
rs769163736 Chr.7: 100091537-100091537 on Build GRCh38
rs772178614 Chr.7: 100091454-100091454 on Build GRCh38
rs768730926 Chr.7: 100091619-100091619 on Build GRCh38
rs753640097 Chr.7: 100091609-100091609 on Build GRCh38
rs200193790 Chr.7: 100091496-100091496 on Build GRCh38
rs200743570 Chr.7: 100091645-100091645 on Build GRCh38
rs371103105 Chr.7: 100091644-100091644 on Build GRCh38
rs557095125 Chr.7: 100091641-100091641 on Build GRCh38
rs745924412 Chr.7: 100091445-100091445 on Build GRCh38
rs1045394766 Chr.7: 100091573-100091573 on Build GRCh38
rs754888968 Chr.7: 100091503-100091503 on Build GRCh38
rs758547231 Chr.7: 100091612-100091612 on Build GRCh38
rs953919313 Chr.7: 100091635-100091635 on Build GRCh38
rs1029010152 Chr.7: 100091632-100091632 on Build GRCh38
rs751530109 Chr.7: 100091499-100091499 on Build GRCh38
rs1011548794 Chr.7: 100091597-100091597 on Build GRCh38
rs901953261 Chr.7: 100091613-100091613 on Build GRCh38
rs920409729 Chr.7: 100091538-100091538 on Build GRCh38
rs769052117 Chr.7: 100091459-100091459 on Build GRCh38
rs748673921 Chr.7: 100091631-100091631 on Build GRCh38
rs188414718 Chr.7: 100091594-100091594 on Build GRCh38
rs758574948 Chr.7: 100091568-100091568 on Build GRCh38
rs931855437 Chr.7: 100091547-100091547 on Build GRCh38
rs767362885 Chr.7: 100091520-100091520 on Build GRCh38
rs893305363 Chr.7: 100091604-100091604 on Build GRCh38
rs61648592 Chr.7: 100091548-100091548 on Build GRCh38
rs758455552 Chr.7: 100091527-100091527 on Build GRCh38
rs762118429 Chr.7: 100091461-100091461 on Build GRCh38
rs1052012443 Chr.7: 100091579-100091579 on Build GRCh38
rs755909532 Chr.7: 100091522-100091522 on Build GRCh38
rs776983712 Chr.7: 100091460-100091460 on Build GRCh38
rs1010437902 Chr.7: 100091605-100091605 on Build GRCh38
rs747607635 Chr.7: 100091458-100091458 on Build GRCh38
rs775968450 Chr.7: 100091457-100091457 on Build GRCh38
rs780282875 Chr.7: 100091614-100091614 on Build GRCh38
rs747064885 Chr.7: 100091618-100091618 on Build GRCh38
rs141560742 Chr.7: 100091608-100091608 on Build GRCh38
rs376648875 Chr.7: 100091535-100091535 on Build GRCh38
rs372933685 Chr.7: 100091571-100091571 on Build GRCh38
rs376848627 Chr.7: 100091554-100091554 on Build GRCh38
rs763076174 Chr.7: 100091639-100091642 on Build GRCh38
rs113017856 Chr.7: 100091536-100091536 on Build GRCh38
rs746155068 Chr.7: 100091526-100091526 on Build GRCh38

View Full Product Details