Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77931471-77931712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536985.1 XM_017021474.1 XM_017021475.1 XM_017021476.1 XM_017021477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526387 c.1188G>A p.A396A Chr.14: 77931703-77931703 on Build GRCh38
5561029 c.1021A>G p.N341D Chr.14: 77931536-77931536 on Build GRCh38
268598 c.1319G>A p.G440D Chr.14: 77931630-77931630 on Build GRCh38
4052649 c.1220G>A p.R407H Chr.14: 77931531-77931531 on Build GRCh38
4462714 c.1054C>T p.L352F Chr.14: 77931569-77931569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008647448 Chr.14: 77931639-77931639 on Build GRCh38
rs775251025 Chr.14: 77931654-77931659 on Build GRCh38
rs752581267 Chr.14: 77931530-77931530 on Build GRCh38
rs56351872 Chr.14: 77931686-77931686 on Build GRCh38
rs201345422 Chr.14: 77931527-77931527 on Build GRCh38
rs748588347 Chr.14: 77931641-77931641 on Build GRCh38
rs764007785 Chr.14: 77931693-77931693 on Build GRCh38
rs769236669 Chr.14: 77931574-77931574 on Build GRCh38
rs764079575 Chr.14: 77931657-77931657 on Build GRCh38
rs545192306 Chr.14: 77931616-77931616 on Build GRCh38
rs144926474 Chr.14: 77931587-77931587 on Build GRCh38
rs760084948 Chr.14: 77931662-77931663 on Build GRCh38
rs746021046 Chr.14: 77931547-77931547 on Build GRCh38
rs766091569 Chr.14: 77931522-77931522 on Build GRCh38
rs764371737 Chr.14: 77931497-77931497 on Build GRCh38
rs367850250 Chr.14: 77931687-77931687 on Build GRCh38
rs922542252 Chr.14: 77931638-77931638 on Build GRCh38
rs762424531 Chr.14: 77931656-77931656 on Build GRCh38
rs762827456 Chr.14: 77931595-77931595 on Build GRCh38
rs143522447 Chr.14: 77931546-77931546 on Build GRCh38
rs141847250 Chr.14: 77931662-77931662 on Build GRCh38
rs773566727 Chr.14: 77931646-77931646 on Build GRCh38
rs111650050 Chr.14: 77931586-77931586 on Build GRCh38
rs904867171 Chr.14: 77931677-77931677 on Build GRCh38
rs781463871 Chr.14: 77931623-77931623 on Build GRCh38
rs746764117 Chr.14: 77931540-77931540 on Build GRCh38
rs149764524 Chr.14: 77931640-77931640 on Build GRCh38
rs149882720 Chr.14: 77931585-77931585 on Build GRCh38
rs976705008 Chr.14: 77931636-77931636 on Build GRCh38
rs774900095 Chr.14: 77931655-77931655 on Build GRCh38
rs760427881 Chr.14: 77931500-77931500 on Build GRCh38
rs74065837 Chr.14: 77931563-77931563 on Build GRCh38
rs751408118 Chr.14: 77931617-77931617 on Build GRCh38
rs547081408 Chr.14: 77931557-77931557 on Build GRCh38
rs199757829 Chr.14: 77931626-77931626 on Build GRCh38
rs757036104 Chr.14: 77931620-77931620 on Build GRCh38
rs752199480 Chr.14: 77931593-77931593 on Build GRCh38
rs769762432 Chr.14: 77931554-77931554 on Build GRCh38
rs139959248 Chr.14: 77931538-77931538 on Build GRCh38
rs147126229 Chr.14: 77931689-77931689 on Build GRCh38
rs762239164 Chr.14: 77931581-77931581 on Build GRCh38
rs759129763 Chr.14: 77931651-77931651 on Build GRCh38
rs540439929 Chr.14: 77931661-77931661 on Build GRCh38
rs140999497 Chr.14: 77931630-77931630 on Build GRCh38
rs774227675 Chr.14: 77931492-77931492 on Build GRCh38
rs757507916 Chr.14: 77931545-77931545 on Build GRCh38
rs112630340 Chr.14: 77931588-77931588 on Build GRCh38
rs750618689 Chr.14: 77931625-77931625 on Build GRCh38
rs905422403 Chr.14: 77931567-77931567 on Build GRCh38
rs139355965 Chr.14: 77931580-77931580 on Build GRCh38
rs753446949 Chr.14: 77931523-77931523 on Build GRCh38
rs772569167 Chr.14: 77931643-77931643 on Build GRCh38
rs999878942 Chr.14: 77931602-77931602 on Build GRCh38
rs765285872 Chr.14: 77931528-77931528 on Build GRCh38
rs758294837 Chr.14: 77931531-77931531 on Build GRCh38
rs920624545 Chr.14: 77931556-77931556 on Build GRCh38
rs749350658 Chr.14: 77931627-77931627 on Build GRCh38
rs961891343 Chr.14: 77931653-77931653 on Build GRCh38
rs771588615 Chr.14: 77931626-77931626 on Build GRCh38
rs750185239 Chr.14: 77931685-77931685 on Build GRCh38
rs767316774 Chr.14: 77931682-77931682 on Build GRCh38
rs1043980378 Chr.14: 77931594-77931594 on Build GRCh38
rs904079150 Chr.14: 77931597-77931597 on Build GRCh38
rs745684727 Chr.14: 77931549-77931549 on Build GRCh38

View Full Product Details