Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58096789-58097050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
UniGene
Hs.26766
Gene Chromosome Location
Chr.14: 57993536-58152305 on Build GRCh38
Transcripts
NM_001001872.3 XM_011536471.2 XM_017021029.1 XM_017021030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6849288 c.1892A>G p.E631G Chr.14: 58096921-58096921 on Build GRCh38
6875897 c.1855G>A p.E619K Chr.14: 58096958-58096958 on Build GRCh38
4778703 c.1858G>T p.D620Y Chr.14: 58096955-58096955 on Build GRCh38
7149267 c.1840G>C p.E614Q Chr.14: 58096973-58096973 on Build GRCh38
470053 c.1832A>G p.E611G Chr.14: 58096981-58096981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780134523 Chr.14: 58096875-58096875 on Build GRCh38
rs766418911 Chr.14: 58096975-58096975 on Build GRCh38
rs762095398 Chr.14: 58096949-58096949 on Build GRCh38
rs755922478 Chr.14: 58097004-58097004 on Build GRCh38
rs754863591 Chr.14: 58096933-58096938 on Build GRCh38
rs759142103 Chr.14: 58096839-58096839 on Build GRCh38
rs747218089 Chr.14: 58096880-58096880 on Build GRCh38
rs867854070 Chr.14: 58096979-58096979 on Build GRCh38
rs754199404 Chr.14: 58096850-58096850 on Build GRCh38
rs769902115 Chr.14: 58096817-58096817 on Build GRCh38
rs781337594 Chr.14: 58096987-58096987 on Build GRCh38
rs752431315 Chr.14: 58096945-58096947 on Build GRCh38
rs764485521 Chr.14: 58096849-58096849 on Build GRCh38
rs767129929 Chr.14: 58096945-58096945 on Build GRCh38
rs758418437 Chr.14: 58096866-58096866 on Build GRCh38
rs762620810 Chr.14: 58096820-58096820 on Build GRCh38
rs762137652 Chr.14: 58096918-58096920 on Build GRCh38
rs62640395 Chr.14: 58096953-58096953 on Build GRCh38
rs781019905 Chr.14: 58096888-58096888 on Build GRCh38
rs750884160 Chr.14: 58096965-58096965 on Build GRCh38
rs755215067 Chr.14: 58096980-58096980 on Build GRCh38
rs780310947 Chr.14: 58096818-58096818 on Build GRCh38
rs768802446 Chr.14: 58096887-58096887 on Build GRCh38
rs751207509 Chr.14: 58096834-58096834 on Build GRCh38
rs564454782 Chr.14: 58096944-58096944 on Build GRCh38
rs1049337788 Chr.14: 58097000-58097000 on Build GRCh38
rs773772686 Chr.14: 58096932-58096932 on Build GRCh38
rs371656172 Chr.14: 58096941-58096955 on Build GRCh38
rs765207548 Chr.14: 58096927-58096929 on Build GRCh38
rs371827034 Chr.14: 58097018-58097018 on Build GRCh38
rs199528318 Chr.14: 58097007-58097007 on Build GRCh38
rs942407626 Chr.14: 58096882-58096882 on Build GRCh38
rs376833564 Chr.14: 58096947-58096947 on Build GRCh38
rs747954144 Chr.14: 58096920-58096920 on Build GRCh38
rs750570835 Chr.14: 58096930-58096944 on Build GRCh38
rs758938973 Chr.14: 58096966-58096966 on Build GRCh38
rs777028058 Chr.14: 58096903-58096914 on Build GRCh38
rs759201414 Chr.14: 58096939-58096939 on Build GRCh38
rs147449063 Chr.14: 58096861-58096861 on Build GRCh38
rs769851282 Chr.14: 58096921-58096921 on Build GRCh38
rs2273442 Chr.14: 58096976-58096976 on Build GRCh38
rs748408929 Chr.14: 58097001-58097001 on Build GRCh38
rs767443274 Chr.14: 58096944-58096945 on Build GRCh38
rs755924926 Chr.14: 58096948-58096956 on Build GRCh38
rs140057603 Chr.14: 58096924-58096924 on Build GRCh38
rs765998834 Chr.14: 58096825-58096825 on Build GRCh38
rs372192668 Chr.14: 58096928-58096928 on Build GRCh38
rs757767365 Chr.14: 58096860-58096860 on Build GRCh38

View Full Product Details