Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11201832-11202071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK6
Entrez Gene ID
57572
Gene Name
dedicator of cytokinesis 6
Gene Aliases
AOS2, ZIR1
UniGene
Hs.591002
Gene Chromosome Location
Chr.19: 11199293-11262502 on Build GRCh38
Transcripts
NM_020812.3 XM_005260000.2 XM_005260001.2 XM_006722804.3 XM_011528150.1 XM_011528151.1 XM_011528152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679056 c.5534G>A p.R1845H Chr.19: 11202049-11202049 on Build GRCh38
3742725 c.5592A>G p.A1864A Chr.19: 11201991-11201991 on Build GRCh38
5017711 c.5543G>C p.G1848A Chr.19: 11202040-11202040 on Build GRCh38
1325513 c.5646C>T p.H1882H Chr.19: 11201937-11201937 on Build GRCh38
2814077 c.5549G>A p.R1850H Chr.19: 11202034-11202034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014330241 Chr.19: 11201919-11201919 on Build GRCh38
rs189395618 Chr.19: 11201963-11201963 on Build GRCh38
rs756548269 Chr.19: 11202015-11202015 on Build GRCh38
rs145128169 Chr.19: 11202002-11202002 on Build GRCh38
rs770042344 Chr.19: 11201906-11201906 on Build GRCh38
rs760235529 Chr.19: 11201932-11201932 on Build GRCh38
rs372223901 Chr.19: 11201993-11201993 on Build GRCh38
rs372106818 Chr.19: 11202003-11202003 on Build GRCh38
rs992035587 Chr.19: 11201999-11201999 on Build GRCh38
rs760209633 Chr.19: 11201976-11201976 on Build GRCh38
rs756208935 Chr.19: 11201962-11201962 on Build GRCh38
rs369620993 Chr.19: 11202010-11202010 on Build GRCh38
rs375177119 Chr.19: 11202007-11202007 on Build GRCh38
rs771893047 Chr.19: 11201927-11201927 on Build GRCh38
rs763644777 Chr.19: 11201935-11201935 on Build GRCh38
rs753162763 Chr.19: 11201960-11201960 on Build GRCh38
rs573654195 Chr.19: 11201996-11201996 on Build GRCh38
rs374323288 Chr.19: 11202039-11202039 on Build GRCh38
rs368166109 Chr.19: 11202049-11202049 on Build GRCh38
rs926739290 Chr.19: 11202044-11202044 on Build GRCh38
rs377629249 Chr.19: 11202027-11202027 on Build GRCh38
rs539660006 Chr.19: 11201897-11201897 on Build GRCh38
rs750861511 Chr.19: 11201861-11201861 on Build GRCh38
rs759587834 Chr.19: 11201926-11201926 on Build GRCh38
rs777898007 Chr.19: 11202016-11202016 on Build GRCh38
rs566803658 Chr.19: 11201923-11201923 on Build GRCh38
rs374242691 Chr.19: 11202012-11202012 on Build GRCh38
rs775119860 Chr.19: 11201931-11201931 on Build GRCh38
rs773952355 Chr.19: 11201925-11201925 on Build GRCh38
rs746066363 Chr.19: 11202035-11202035 on Build GRCh38
rs137932140 Chr.19: 11202025-11202025 on Build GRCh38
rs201858507 Chr.19: 11202034-11202034 on Build GRCh38
rs776328092 Chr.19: 11202050-11202050 on Build GRCh38
rs573580688 Chr.19: 11201988-11201988 on Build GRCh38
rs551574920 Chr.19: 11201909-11201909 on Build GRCh38
rs755557243 Chr.19: 11201955-11201955 on Build GRCh38
rs537400350 Chr.19: 11201992-11201992 on Build GRCh38
rs376993154 Chr.19: 11202040-11202040 on Build GRCh38
rs758853300 Chr.19: 11201864-11201864 on Build GRCh38
rs747006582 Chr.19: 11202048-11202048 on Build GRCh38
rs368573184 Chr.19: 11201936-11201936 on Build GRCh38
rs115035890 Chr.19: 11201995-11201995 on Build GRCh38
rs374250039 Chr.19: 11201956-11201956 on Build GRCh38
rs200959822 Chr.19: 11201937-11201937 on Build GRCh38
rs780222506 Chr.19: 11201865-11201865 on Build GRCh38
rs771745764 Chr.19: 11201942-11201942 on Build GRCh38
rs774273979 Chr.19: 11201896-11201896 on Build GRCh38
rs773454770 Chr.19: 11201908-11201908 on Build GRCh38
rs781389396 Chr.19: 11201884-11201884 on Build GRCh38
rs373843420 Chr.19: 11201880-11201880 on Build GRCh38
rs768282386 Chr.19: 11201994-11201994 on Build GRCh38
rs892446076 Chr.19: 11202046-11202046 on Build GRCh38
rs1028777050 Chr.19: 11201971-11201971 on Build GRCh38
rs201873244 Chr.19: 11201975-11201975 on Build GRCh38
rs964141123 Chr.19: 11201943-11201943 on Build GRCh38
rs200790813 Chr.19: 11201859-11201859 on Build GRCh38
rs769306856 Chr.19: 11202051-11202051 on Build GRCh38

View Full Product Details