Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197512253-197512502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
UniGene
Hs.274539
Gene Chromosome Location
Chr.3: 197509783-197573323 on Build GRCh38
Transcripts
NM_004051.4 NM_203314.2 NM_203315.2 XM_005269352.3 XM_005269355.4 XM_011513067.2 XM_017007007.1 XM_017007008.1 XM_017007009.1 XM_017007010.1 XM_017007011.1 XM_017007012.1 XM_017007014.1 XM_017007015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6164166 c.641G>T p.G214V Chr.3: 197512286-197512286 on Build GRCh38
1566837 c.620C>T p.P207L Chr.3: 197512307-197512307 on Build GRCh38
7272579 c.613C>T p.R205C Chr.3: 197512314-197512314 on Build GRCh38
3660457 c.609G>T p.P203P Chr.3: 197512318-197512318 on Build GRCh38
4152396 c.589C>G p.L197V Chr.3: 197512338-197512338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771675343 Chr.3: 197512378-197512378 on Build GRCh38
rs776427253 Chr.3: 197512396-197512396 on Build GRCh38
rs9325394 Chr.3: 197512395-197512395 on Build GRCh38
rs373897141 Chr.3: 197512368-197512368 on Build GRCh38
rs774160942 Chr.3: 197512318-197512318 on Build GRCh38
rs752915471 Chr.3: 197512359-197512359 on Build GRCh38
rs200605471 Chr.3: 197512307-197512307 on Build GRCh38
rs1039817202 Chr.3: 197512418-197512418 on Build GRCh38
rs763236268 Chr.3: 197512339-197512339 on Build GRCh38
rs767131775 Chr.3: 197512271-197512271 on Build GRCh38
rs559038515 Chr.3: 197512273-197512273 on Build GRCh38
rs1050813950 Chr.3: 197512373-197512373 on Build GRCh38
rs755033433 Chr.3: 197512357-197512357 on Build GRCh38
rs1045820669 Chr.3: 197512455-197512455 on Build GRCh38
rs964914209 Chr.3: 197512343-197512343 on Build GRCh38
rs752081360 Chr.3: 197512406-197512406 on Build GRCh38
rs760442245 Chr.3: 197512331-197512331 on Build GRCh38
rs920367550 Chr.3: 197512362-197512362 on Build GRCh38
rs200618218 Chr.3: 197512342-197512342 on Build GRCh38
rs141633234 Chr.3: 197512363-197512363 on Build GRCh38
rs368597951 Chr.3: 197512314-197512314 on Build GRCh38
rs757649832 Chr.3: 197512277-197512277 on Build GRCh38
rs917233468 Chr.3: 197512294-197512294 on Build GRCh38
rs375263576 Chr.3: 197512301-197512301 on Build GRCh38
rs1001200935 Chr.3: 197512456-197512456 on Build GRCh38
rs766839210 Chr.3: 197512353-197512353 on Build GRCh38
rs766675169 Chr.3: 197512404-197512404 on Build GRCh38
rs140269063 Chr.3: 197512319-197512319 on Build GRCh38
rs371927738 Chr.3: 197512270-197512270 on Build GRCh38
rs750875619 Chr.3: 197512338-197512338 on Build GRCh38
rs779985174 Chr.3: 197512288-197512288 on Build GRCh38
rs137863421 Chr.3: 197512361-197512361 on Build GRCh38
rs773061706 Chr.3: 197512312-197512312 on Build GRCh38
rs771506759 Chr.3: 197512324-197512324 on Build GRCh38
rs776226119 Chr.3: 197512479-197512479 on Build GRCh38
rs746587221 Chr.3: 197512433-197512433 on Build GRCh38
rs114484832 Chr.3: 197512306-197512306 on Build GRCh38
rs114600793 Chr.3: 197512284-197512284 on Build GRCh38
rs150101930 Chr.3: 197512386-197512386 on Build GRCh38
rs144830970 Chr.3: 197512286-197512286 on Build GRCh38
rs752035471 Chr.3: 197512356-197512356 on Build GRCh38
rs765743905 Chr.3: 197512278-197512278 on Build GRCh38
rs763336760 Chr.3: 197512398-197512398 on Build GRCh38
rs927563864 Chr.3: 197512322-197512322 on Build GRCh38
rs749172620 Chr.3: 197512313-197512313 on Build GRCh38
rs745845198 Chr.3: 197512377-197512377 on Build GRCh38
rs903762297 Chr.3: 197512422-197512422 on Build GRCh38
rs779465065 Chr.3: 197512425-197512425 on Build GRCh38
rs755013846 Chr.3: 197512293-197512293 on Build GRCh38
rs747048497 Chr.3: 197512292-197512292 on Build GRCh38
rs768290470 Chr.3: 197512434-197512434 on Build GRCh38
rs775259877 Chr.3: 197512329-197512329 on Build GRCh38
rs542445174 Chr.3: 197512332-197512332 on Build GRCh38
rs143508627 Chr.3: 197512360-197512360 on Build GRCh38

View Full Product Details