Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 35381841-35382114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH6
Entrez Gene ID
89978
Gene Name
diphthamine biosynthesis 6
Gene Aliases
ATPBD4
UniGene
Hs.107196
Gene Chromosome Location
Chr.15: 35217356-35546222 on Build GRCh38
Transcripts
NM_080650.3 XM_011522160.1 XM_017022707.1 XM_017022708.1 XM_017022709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1707988 c.568C>T p.L190F Chr.15: 35381916-35381916 on Build GRCh38
7205856 c.585A>G p.G195G Chr.15: 35381899-35381899 on Build GRCh38
6648857 c.633T>A p.D211E Chr.15: 35381851-35381851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953949304 Chr.15: 35381880-35381880 on Build GRCh38
rs7497080 Chr.15: 35381935-35381935 on Build GRCh38
rs763186764 Chr.15: 35381888-35381888 on Build GRCh38
rs760915793 Chr.15: 35381924-35381924 on Build GRCh38
rs1014129161 Chr.15: 35381988-35381988 on Build GRCh38
rs376543379 Chr.15: 35381951-35381951 on Build GRCh38
rs753311818 Chr.15: 35381908-35381908 on Build GRCh38
rs879760657 Chr.15: 35382053-35382053 on Build GRCh38
rs200564485 Chr.15: 35381878-35381878 on Build GRCh38
rs948932064 Chr.15: 35382037-35382037 on Build GRCh38
rs186793838 Chr.15: 35382024-35382024 on Build GRCh38
rs773401816 Chr.15: 35381885-35381885 on Build GRCh38
rs758151005 Chr.15: 35381933-35381933 on Build GRCh38
rs935914474 Chr.15: 35381962-35381962 on Build GRCh38
rs769674669 Chr.15: 35381883-35381883 on Build GRCh38
rs189618694 Chr.15: 35381927-35381927 on Build GRCh38
rs905779583 Chr.15: 35381994-35381994 on Build GRCh38
rs756823738 Chr.15: 35382016-35382016 on Build GRCh38
rs80019930 Chr.15: 35381983-35381983 on Build GRCh38
rs369180853 Chr.15: 35381944-35381944 on Build GRCh38
rs905955878 Chr.15: 35382000-35382000 on Build GRCh38
rs866750356 Chr.15: 35381889-35381889 on Build GRCh38
rs760030551 Chr.15: 35381903-35381903 on Build GRCh38
rs774419259 Chr.15: 35381902-35381902 on Build GRCh38
rs779636291 Chr.15: 35381934-35381934 on Build GRCh38
rs149996784 Chr.15: 35381905-35381905 on Build GRCh38
rs962905533 Chr.15: 35382046-35382046 on Build GRCh38
rs781253434 Chr.15: 35381872-35381872 on Build GRCh38
rs763908910 Chr.15: 35381934-35381935 on Build GRCh38
rs151276162 Chr.15: 35382002-35382002 on Build GRCh38
rs1024805182 Chr.15: 35381992-35381992 on Build GRCh38
rs1044123224 Chr.15: 35382048-35382048 on Build GRCh38
rs182209664 Chr.15: 35381997-35381997 on Build GRCh38
rs750045657 Chr.15: 35381929-35381929 on Build GRCh38
rs770958547 Chr.15: 35381900-35381900 on Build GRCh38
rs191702569 Chr.15: 35382049-35382049 on Build GRCh38

View Full Product Details