Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230705764-230706029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGT
Entrez Gene ID
183
Gene Name
angiotensinogen
Gene Aliases
ANHU, SERPINA8
UniGene
Hs.19383
Gene Chromosome Location
Chr.1: 230702523-230714590 on Build GRCh38
Transcripts
NM_000029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596480 c.1120C>T p.P374S Chr.1: 230705937-230705937 on Build GRCh38
4521416 c.1110G>A p.K370K Chr.1: 230705947-230705947 on Build GRCh38
6283385 c.1048G>T p.A350S Chr.1: 230706009-230706009 on Build GRCh38
905373 c.1110G>T p.K370N Chr.1: 230705947-230705947 on Build GRCh38
7167108 c.1051T>C p.S351P Chr.1: 230706006-230706006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11568042 Chr.1: 230705815-230705815 on Build GRCh38
rs11568043 Chr.1: 230705805-230705805 on Build GRCh38
rs386640205 Chr.1: 230705813-230705815 on Build GRCh38
rs1042496077 Chr.1: 230705905-230705905 on Build GRCh38
rs775681269 Chr.1: 230705892-230705892 on Build GRCh38
rs778806374 Chr.1: 230705970-230705970 on Build GRCh38
rs868602913 Chr.1: 230705953-230705953 on Build GRCh38
rs773199166 Chr.1: 230705892-230705892 on Build GRCh38
rs757457429 Chr.1: 230705968-230705968 on Build GRCh38
rs933166266 Chr.1: 230705828-230705828 on Build GRCh38
rs764728122 Chr.1: 230705920-230705920 on Build GRCh38
rs929340143 Chr.1: 230705814-230705814 on Build GRCh38
rs547944063 Chr.1: 230705790-230705790 on Build GRCh38
rs200520566 Chr.1: 230705919-230705919 on Build GRCh38
rs371295393 Chr.1: 230705924-230705924 on Build GRCh38
rs761251154 Chr.1: 230705894-230705894 on Build GRCh38
rs115636911 Chr.1: 230705925-230705925 on Build GRCh38
rs878964672 Chr.1: 230705803-230705803 on Build GRCh38
rs1012870153 Chr.1: 230705935-230705935 on Build GRCh38
rs1032008704 Chr.1: 230705873-230705873 on Build GRCh38
rs7080 Chr.1: 230705941-230705941 on Build GRCh38
rs149690396 Chr.1: 230705883-230705883 on Build GRCh38
rs61757179 Chr.1: 230705885-230705885 on Build GRCh38
rs61757181 Chr.1: 230705791-230705791 on Build GRCh38
rs900292240 Chr.1: 230705816-230705816 on Build GRCh38
rs61762528 Chr.1: 230705871-230705871 on Build GRCh38
rs555708850 Chr.1: 230705870-230705870 on Build GRCh38
rs1040581763 Chr.1: 230705866-230705866 on Build GRCh38
rs902950394 Chr.1: 230705916-230705916 on Build GRCh38
rs566689024 Chr.1: 230705853-230705853 on Build GRCh38
rs984378489 Chr.1: 230705997-230705997 on Build GRCh38
rs758798996 Chr.1: 230705804-230705804 on Build GRCh38
rs111650082 Chr.1: 230705797-230705797 on Build GRCh38
rs557343839 Chr.1: 230705826-230705826 on Build GRCh38
rs376668080 Chr.1: 230706007-230706007 on Build GRCh38
rs61757180 Chr.1: 230705865-230705865 on Build GRCh38
rs746136385 Chr.1: 230705984-230705984 on Build GRCh38
rs74315283 Chr.1: 230705933-230705933 on Build GRCh38
rs772512439 Chr.1: 230705890-230705890 on Build GRCh38
rs201162475 Chr.1: 230705934-230705934 on Build GRCh38
rs61762529 Chr.1: 230706000-230706000 on Build GRCh38
rs150452789 Chr.1: 230706003-230706003 on Build GRCh38
rs772123904 Chr.1: 230705994-230705994 on Build GRCh38
rs3827749 Chr.1: 230705813-230705813 on Build GRCh38
rs777921457 Chr.1: 230705937-230705937 on Build GRCh38
rs530553482 Chr.1: 230705796-230705796 on Build GRCh38
rs894014191 Chr.1: 230705875-230705875 on Build GRCh38
rs570157525 Chr.1: 230705812-230705812 on Build GRCh38

View Full Product Details