Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89603080-89603337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B1
Entrez Gene ID
490
Gene Name
ATPase plasma membrane Ca2+ transporting 1
Gene Aliases
PMCA1, PMCA1kb
UniGene
Hs.506276
Gene Chromosome Location
Chr.12: 89588049-89709353 on Build GRCh38
Transcripts
NM_001001323.1 NM_001682.2 XM_011538407.2 XM_017019357.1 XM_017019358.1 XM_017019359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
431963 c.3007A>G p.I1003V Chr.12: 89603096-89603096 on Build GRCh38
6607592 c.2958C>T p.N986N Chr.12: 89603145-89603145 on Build GRCh38
5587247 c.3001G>A p.E1001K Chr.12: 89603102-89603102 on Build GRCh38
3772746 c.2971C>T p.R991W Chr.12: 89603132-89603132 on Build GRCh38
6355759 c.2959G>A p.E987K Chr.12: 89603144-89603144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780919995 Chr.12: 89603248-89603248 on Build GRCh38
rs779822283 Chr.12: 89603218-89603218 on Build GRCh38
rs771630385 Chr.12: 89603296-89603296 on Build GRCh38
rs370810713 Chr.12: 89603165-89603165 on Build GRCh38
rs11105341 Chr.12: 89603301-89603301 on Build GRCh38
rs377272508 Chr.12: 89603125-89603125 on Build GRCh38
rs778675473 Chr.12: 89603204-89603204 on Build GRCh38
rs376290022 Chr.12: 89603269-89603269 on Build GRCh38
rs756240515 Chr.12: 89603187-89603187 on Build GRCh38
rs1005125190 Chr.12: 89603145-89603145 on Build GRCh38
rs374196525 Chr.12: 89603281-89603281 on Build GRCh38
rs966303879 Chr.12: 89603226-89603226 on Build GRCh38
rs752693675 Chr.12: 89603180-89603180 on Build GRCh38
rs746847972 Chr.12: 89603237-89603237 on Build GRCh38
rs758129782 Chr.12: 89603208-89603208 on Build GRCh38
rs189003061 Chr.12: 89603241-89603241 on Build GRCh38
rs750364043 Chr.12: 89603207-89603207 on Build GRCh38
rs370788275 Chr.12: 89603276-89603276 on Build GRCh38
rs763543000 Chr.12: 89603288-89603288 on Build GRCh38
rs767818173 Chr.12: 89603127-89603127 on Build GRCh38
rs1015210218 Chr.12: 89603196-89603196 on Build GRCh38

View Full Product Details