Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2189533-2189792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOT1L
Entrez Gene ID
84444
Gene Name
DOT1 like histone lysine methyltransferase
Gene Aliases
DOT1, KMT4
UniGene
Hs.713641
Gene Chromosome Location
Chr.19: 2163963-2232578 on Build GRCh38
Transcripts
NM_032482.2 XM_005259659.3 XM_005259660.3 XM_011528359.2 XM_011528360.1 XM_017027366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1589919 c.202T>C p.F68L Chr.19: 2189733-2189733 on Build GRCh38
7244135 c.219G>A p.R73R Chr.19: 2189750-2189750 on Build GRCh38
6648709 c.225C>T p.C75C Chr.19: 2189756-2189756 on Build GRCh38
993706 c.202T>C p.F68L Chr.19: 2189733-2189733 on Build GRCh38
3103069 c.204C>T p.F68F Chr.19: 2189735-2189735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775691456 Chr.19: 2189685-2189685 on Build GRCh38
rs199909162 Chr.19: 2189756-2189756 on Build GRCh38
rs537079884 Chr.19: 2189678-2189678 on Build GRCh38
rs759089252 Chr.19: 2189702-2189702 on Build GRCh38
rs368740625 Chr.19: 2189707-2189707 on Build GRCh38
rs764139743 Chr.19: 2189716-2189716 on Build GRCh38
rs913160679 Chr.19: 2189665-2189665 on Build GRCh38
rs923095008 Chr.19: 2189573-2189573 on Build GRCh38
rs931656606 Chr.19: 2189657-2189657 on Build GRCh38
rs988728618 Chr.19: 2189582-2189582 on Build GRCh38
rs757442864 Chr.19: 2189723-2189723 on Build GRCh38
rs370257976 Chr.19: 2189697-2189697 on Build GRCh38
rs907769446 Chr.19: 2189579-2189579 on Build GRCh38
rs1043460223 Chr.19: 2189689-2189689 on Build GRCh38
rs375845994 Chr.19: 2189629-2189629 on Build GRCh38
rs888708392 Chr.19: 2189724-2189724 on Build GRCh38
rs150316325 Chr.19: 2189750-2189750 on Build GRCh38
rs779102020 Chr.19: 2189729-2189729 on Build GRCh38
rs372161368 Chr.19: 2189735-2189735 on Build GRCh38
rs755870956 Chr.19: 2189711-2189711 on Build GRCh38
rs555421971 Chr.19: 2189681-2189681 on Build GRCh38
rs973089628 Chr.19: 2189612-2189612 on Build GRCh38
rs1023736894 Chr.19: 2189565-2189565 on Build GRCh38
rs750379407 Chr.19: 2189576-2189576 on Build GRCh38
rs753779025 Chr.19: 2189717-2189717 on Build GRCh38
rs769020592 Chr.19: 2189765-2189765 on Build GRCh38
rs758532148 Chr.19: 2189736-2189736 on Build GRCh38
rs1050112814 Chr.19: 2189677-2189677 on Build GRCh38
rs367786294 Chr.19: 2189700-2189700 on Build GRCh38
rs920214702 Chr.19: 2189632-2189632 on Build GRCh38
rs569662365 Chr.19: 2189592-2189592 on Build GRCh38
rs551749263 Chr.19: 2189571-2189571 on Build GRCh38
rs762475482 Chr.19: 2189698-2189698 on Build GRCh38
rs761298039 Chr.19: 2189687-2189687 on Build GRCh38
rs186572260 Chr.19: 2189699-2189699 on Build GRCh38
rs769051965 Chr.19: 2189691-2189691 on Build GRCh38
rs1041233088 Chr.19: 2189695-2189695 on Build GRCh38

View Full Product Details