Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178865588-178865837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL1
Entrez Gene ID
9068
Gene Name
angiopoietin like 1
Gene Aliases
ANG3, ANGPT3, ARP1, AngY, UNQ162, dJ595C2.2
UniGene
Hs.591474
Gene Chromosome Location
Chr.1: 178849535-178871353 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

Gene Symbol
RALGPS2
Entrez Gene ID
55103
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Gene Aliases
dJ595C2.1
UniGene
Hs.632485
Gene Chromosome Location
Chr.1: 178725147-178921842 on Build GRCh38
Transcripts
NM_004673.3 XM_005245577.2 NM_001286247.1 NM_152663.4 XM_006711410.3 XM_006711411.3 XM_011509688.1 XM_017001591.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6123034 c.62G>T p.C21F Chr.1: 178865715-178865715 on Build GRCh38
3671516 c.40C>A p.L14I Chr.1: 178865737-178865737 on Build GRCh38
1668240 c.143A>C p.K48T Chr.1: 178865634-178865634 on Build GRCh38
5477824 c.13A>G p.T5A Chr.1: 178865764-178865764 on Build GRCh38
2094912 c.50A>G p.D17G Chr.1: 178865727-178865727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540728063 Chr.1: 178865665-178865665 on Build GRCh38
rs141156731 Chr.1: 178865718-178865718 on Build GRCh38
rs377060114 Chr.1: 178865620-178865620 on Build GRCh38
rs970510326 Chr.1: 178865784-178865784 on Build GRCh38
rs749154423 Chr.1: 178865678-178865678 on Build GRCh38
rs777741584 Chr.1: 178865803-178865803 on Build GRCh38
rs779566235 Chr.1: 178865669-178865669 on Build GRCh38
rs918375801 Chr.1: 178865801-178865801 on Build GRCh38
rs561838589 Chr.1: 178865732-178865732 on Build GRCh38
rs761000586 Chr.1: 178865716-178865716 on Build GRCh38
rs757024564 Chr.1: 178865745-178865748 on Build GRCh38
rs148415335 Chr.1: 178865664-178865664 on Build GRCh38
rs773283978 Chr.1: 178865707-178865707 on Build GRCh38
rs577502486 Chr.1: 178865635-178865635 on Build GRCh38
rs183060436 Chr.1: 178865706-178865706 on Build GRCh38
rs972558764 Chr.1: 178865788-178865788 on Build GRCh38
rs760804077 Chr.1: 178865613-178865613 on Build GRCh38
rs780706960 Chr.1: 178865660-178865660 on Build GRCh38
rs142483649 Chr.1: 178865632-178865632 on Build GRCh38
rs147201688 Chr.1: 178865734-178865734 on Build GRCh38
rs779478305 Chr.1: 178865775-178865775 on Build GRCh38
rs575220046 Chr.1: 178865787-178865787 on Build GRCh38
rs368890927 Chr.1: 178865709-178865709 on Build GRCh38
rs764571745 Chr.1: 178865764-178865764 on Build GRCh38
rs779013092 Chr.1: 178865797-178865797 on Build GRCh38
rs765650889 Chr.1: 178865747-178865747 on Build GRCh38
rs753315982 Chr.1: 178865757-178865757 on Build GRCh38
rs754347008 Chr.1: 178865627-178865627 on Build GRCh38
rs953959557 Chr.1: 178865708-178865708 on Build GRCh38
rs771715085 Chr.1: 178865798-178865798 on Build GRCh38
rs1001307337 Chr.1: 178865724-178865724 on Build GRCh38
rs144685672 Chr.1: 178865766-178865766 on Build GRCh38
rs569779866 Chr.1: 178865693-178865693 on Build GRCh38
rs755763170 Chr.1: 178865773-178865773 on Build GRCh38
rs373962000 Chr.1: 178865645-178865645 on Build GRCh38
rs753330812 Chr.1: 178865776-178865776 on Build GRCh38
rs747888406 Chr.1: 178865696-178865696 on Build GRCh38
rs758857810 Chr.1: 178865655-178865655 on Build GRCh38
rs754580604 Chr.1: 178865786-178865786 on Build GRCh38
rs748164009 Chr.1: 178865791-178865791 on Build GRCh38

View Full Product Details