Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94969932-94970157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CWC15
Entrez Gene ID
51503
Gene Name
CWC15 spliceosome-associated protein
Gene Aliases
AD002, C11orf5, Cwf15, HSPC148, ORF5
UniGene
Hs.503597
Gene Chromosome Location
Chr.11: 94962622-94973612 on Build GRCh38
Transcripts
NM_016403.3 XM_011542868.2 XM_017017916.1

SNP Detail

SNP ID Chromosome Location
rs587712206 Chr.11: 94970050-94970050 on Build GRCh38
rs782133500 Chr.11: 94969984-94969984 on Build GRCh38
rs909246975 Chr.11: 94970122-94970122 on Build GRCh38
rs782715580 Chr.11: 94970100-94970100 on Build GRCh38
rs781885079 Chr.11: 94970102-94970102 on Build GRCh38
rs782050847 Chr.11: 94970074-94970074 on Build GRCh38
rs374678614 Chr.11: 94970104-94970104 on Build GRCh38
rs781986849 Chr.11: 94969979-94969979 on Build GRCh38
rs180957914 Chr.11: 94969968-94969968 on Build GRCh38
rs781790429 Chr.11: 94969965-94969965 on Build GRCh38
rs782731085 Chr.11: 94970022-94970022 on Build GRCh38

View Full Product Details