Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33277458-33277685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT4
Entrez Gene ID
8705
Gene Name
beta-1,3-galactosyltransferase 4
Gene Aliases
BETA3GALT4, GALT2, GALT4
UniGene
Hs.534375
Gene Chromosome Location
Chr.6: 33277140-33278825 on Build GRCh38
Transcripts
NM_003782.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3349582 c.171G>T p.L57F Chr.6: 33277590-33277590 on Build GRCh38
4855117 c.229G>A p.V77M Chr.6: 33277648-33277648 on Build GRCh38
237038 c.99G>A p.L33L Chr.6: 33277518-33277518 on Build GRCh38
451296 c.218T>G p.L73R Chr.6: 33277637-33277637 on Build GRCh38
115810 c.46C>G p.L16V Chr.6: 33277465-33277465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747459978 Chr.6: 33277621-33277621 on Build GRCh38
rs772747220 Chr.6: 33277612-33277612 on Build GRCh38
rs772898354 Chr.6: 33277500-33277500 on Build GRCh38
rs763825739 Chr.6: 33277662-33277662 on Build GRCh38
rs751142174 Chr.6: 33277621-33277621 on Build GRCh38
rs775999984 Chr.6: 33277624-33277624 on Build GRCh38
rs62639320 Chr.6: 33277576-33277576 on Build GRCh38
rs774515794 Chr.6: 33277483-33277483 on Build GRCh38
rs375171630 Chr.6: 33277507-33277507 on Build GRCh38
rs770478999 Chr.6: 33277623-33277623 on Build GRCh38
rs779392651 Chr.6: 33277607-33277607 on Build GRCh38
rs2231259 Chr.6: 33277578-33277578 on Build GRCh38
rs370873982 Chr.6: 33277485-33277485 on Build GRCh38
rs764689356 Chr.6: 33277617-33277617 on Build GRCh38
rs755154942 Chr.6: 33277601-33277601 on Build GRCh38
rs758376733 Chr.6: 33277551-33277551 on Build GRCh38
rs779942572 Chr.6: 33277587-33277587 on Build GRCh38
rs759291316 Chr.6: 33277626-33277626 on Build GRCh38
rs760233433 Chr.6: 33277501-33277501 on Build GRCh38
rs957962678 Chr.6: 33277532-33277532 on Build GRCh38
rs72654491 Chr.6: 33277664-33277664 on Build GRCh38
rs765145081 Chr.6: 33277544-33277544 on Build GRCh38
rs763646603 Chr.6: 33277564-33277564 on Build GRCh38
rs45531637 Chr.6: 33277559-33277559 on Build GRCh38
rs201912153 Chr.6: 33277482-33277482 on Build GRCh38
rs751456996 Chr.6: 33277577-33277577 on Build GRCh38
rs765769178 Chr.6: 33277504-33277504 on Build GRCh38
rs775300073 Chr.6: 33277632-33277632 on Build GRCh38
rs756753916 Chr.6: 33277630-33277630 on Build GRCh38
rs764769925 Chr.6: 33277628-33277628 on Build GRCh38
rs989841952 Chr.6: 33277570-33277570 on Build GRCh38
rs759042116 Chr.6: 33277530-33277530 on Build GRCh38
rs1041315808 Chr.6: 33277481-33277481 on Build GRCh38
rs367923224 Chr.6: 33277609-33277609 on Build GRCh38
rs752417769 Chr.6: 33277548-33277548 on Build GRCh38
rs141784444 Chr.6: 33277620-33277620 on Build GRCh38
rs772693757 Chr.6: 33277492-33277492 on Build GRCh38
rs200023686 Chr.6: 33277636-33277636 on Build GRCh38
rs749533027 Chr.6: 33277599-33277599 on Build GRCh38

View Full Product Details