Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59479217-59479462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L3HYPDH
Entrez Gene ID
112849
Gene Name
trans-L-3-hydroxyproline dehydratase
Gene Aliases
C14orf149
UniGene
Hs.729061
Gene Chromosome Location
Chr.14: 59465545-59484438 on Build GRCh38
Transcripts
NM_144581.1 XM_005267298.3 XM_011536386.2 XM_011536387.1 XM_011536388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276891 c.509-3_509-2delTA p.? Chr.14: 59479353-59479354 on Build GRCh38
6076024 c.551A>G p.D184G Chr.14: 59479309-59479309 on Build GRCh38
956626 c.509A>G p.D170G Chr.14: 59479351-59479351 on Build GRCh38
73892 c.619G>C p.A207P Chr.14: 59479241-59479241 on Build GRCh38
5362549 c.565G>A p.G189S Chr.14: 59479295-59479295 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999397758 Chr.14: 59479397-59479397 on Build GRCh38
rs896707458 Chr.14: 59479254-59479254 on Build GRCh38
rs774372079 Chr.14: 59479277-59479277 on Build GRCh38
rs1752436 Chr.14: 59479381-59479381 on Build GRCh38
rs757614954 Chr.14: 59479392-59479392 on Build GRCh38
rs760391624 Chr.14: 59479293-59479293 on Build GRCh38
rs757223016 Chr.14: 59479299-59479299 on Build GRCh38
rs751192604 Chr.14: 59479382-59479382 on Build GRCh38
rs752960124 Chr.14: 59479292-59479292 on Build GRCh38
rs763829681 Chr.14: 59479353-59479353 on Build GRCh38
rs771295282 Chr.14: 59479374-59479376 on Build GRCh38
rs1011812267 Chr.14: 59479337-59479337 on Build GRCh38
rs376557062 Chr.14: 59479384-59479384 on Build GRCh38
rs775593244 Chr.14: 59479345-59479345 on Build GRCh38
rs778148908 Chr.14: 59479327-59479327 on Build GRCh38
rs570229777 Chr.14: 59479338-59479338 on Build GRCh38
rs749712349 Chr.14: 59479385-59479385 on Build GRCh38
rs757983063 Chr.14: 59479380-59479380 on Build GRCh38
rs376470629 Chr.14: 59479305-59479305 on Build GRCh38
rs753745830 Chr.14: 59479296-59479296 on Build GRCh38
rs753710027 Chr.14: 59479360-59479360 on Build GRCh38
rs774827441 Chr.14: 59479336-59479336 on Build GRCh38
rs570910268 Chr.14: 59479428-59479428 on Build GRCh38
rs760906608 Chr.14: 59479346-59479346 on Build GRCh38
rs754617445 Chr.14: 59479383-59479383 on Build GRCh38
rs773596813 Chr.14: 59479248-59479248 on Build GRCh38
rs757964884 Chr.14: 59479308-59479308 on Build GRCh38
rs770186762 Chr.14: 59479246-59479246 on Build GRCh38
rs771479559 Chr.14: 59479330-59479330 on Build GRCh38
rs751687274 Chr.14: 59479317-59479317 on Build GRCh38
rs763459368 Chr.14: 59479263-59479263 on Build GRCh38
rs749998968 Chr.14: 59479306-59479306 on Build GRCh38
rs765319378 Chr.14: 59479363-59479363 on Build GRCh38
rs772124519 Chr.14: 59479339-59479339 on Build GRCh38
rs770067663 Chr.14: 59479320-59479320 on Build GRCh38
rs779635372 Chr.14: 59479313-59479313 on Build GRCh38
rs759591061 Chr.14: 59479281-59479281 on Build GRCh38
rs111613204 Chr.14: 59479237-59479237 on Build GRCh38
rs774524333 Chr.14: 59479389-59479400 on Build GRCh38
rs1752435 Chr.14: 59479376-59479376 on Build GRCh38
rs749458307 Chr.14: 59479353-59479354 on Build GRCh38
rs767786662 Chr.14: 59479285-59479285 on Build GRCh38
rs771019732 Chr.14: 59479271-59479271 on Build GRCh38
rs370038481 Chr.14: 59479316-59479316 on Build GRCh38
rs1050554079 Chr.14: 59479258-59479258 on Build GRCh38

View Full Product Details