Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98794465-98794738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
UniGene
Hs.469398
Gene Chromosome Location
Chr.2: 98793846-98936221 on Build GRCh38
Transcripts
NM_207362.2 XM_005263923.3 XM_005263924.3 XM_005263926.3 XM_011511093.1 XM_011511094.1 XM_011511095.2 XM_011511096.1 XM_017003971.1 XM_017003972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024277 c.2852C>A p.S951Y Chr.2: 98794569-98794569 on Build GRCh38
3660033 c.2872C>T p.P958S Chr.2: 98794549-98794549 on Build GRCh38
4096554 c.2858C>T p.A953V Chr.2: 98794563-98794563 on Build GRCh38
4484945 c.2875C>T p.Q959* Chr.2: 98794546-98794546 on Build GRCh38
5863065 c.2852C>T p.S951F Chr.2: 98794569-98794569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757788858 Chr.2: 98794577-98794577 on Build GRCh38
rs753171458 Chr.2: 98794529-98794529 on Build GRCh38
rs764574859 Chr.2: 98794704-98794704 on Build GRCh38
rs745408051 Chr.2: 98794575-98794575 on Build GRCh38
rs553168409 Chr.2: 98794616-98794616 on Build GRCh38
rs866676355 Chr.2: 98794683-98794683 on Build GRCh38
rs202214577 Chr.2: 98794514-98794514 on Build GRCh38
rs769848025 Chr.2: 98794496-98794496 on Build GRCh38
rs952252795 Chr.2: 98794590-98794590 on Build GRCh38
rs775556708 Chr.2: 98794640-98794640 on Build GRCh38
rs762727954 Chr.2: 98794499-98794499 on Build GRCh38
rs774086007 Chr.2: 98794656-98794656 on Build GRCh38
rs754637007 Chr.2: 98794697-98794697 on Build GRCh38
rs761755303 Chr.2: 98794670-98794670 on Build GRCh38
rs751471515 Chr.2: 98794503-98794503 on Build GRCh38
rs767293449 Chr.2: 98794673-98794673 on Build GRCh38
rs776379200 Chr.2: 98794519-98794519 on Build GRCh38
rs536508711 Chr.2: 98794517-98794517 on Build GRCh38
rs781342768 Chr.2: 98794596-98794596 on Build GRCh38
rs758814370 Chr.2: 98794557-98794557 on Build GRCh38
rs147783340 Chr.2: 98794688-98794688 on Build GRCh38
rs759824386 Chr.2: 98794495-98794495 on Build GRCh38
rs779569856 Chr.2: 98794527-98794527 on Build GRCh38
rs573076500 Chr.2: 98794693-98794693 on Build GRCh38
rs138898823 Chr.2: 98794505-98794505 on Build GRCh38
rs770117455 Chr.2: 98794628-98794628 on Build GRCh38
rs775357470 Chr.2: 98794497-98794497 on Build GRCh38
rs764073057 Chr.2: 98794500-98794500 on Build GRCh38
rs772847499 Chr.2: 98794678-98794678 on Build GRCh38
rs768474074 Chr.2: 98794653-98794653 on Build GRCh38
rs749125973 Chr.2: 98794650-98794650 on Build GRCh38
rs766104959 Chr.2: 98794682-98794682 on Build GRCh38
rs776532365 Chr.2: 98794493-98794493 on Build GRCh38
rs778464030 Chr.2: 98794570-98794570 on Build GRCh38

View Full Product Details