Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63533345-63533616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB7
Entrez Gene ID
84455
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
-
UniGene
Hs.652324
Gene Chromosome Location
Chr.1: 63523301-63585400 on Build GRCh38
Transcripts
NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846902 c.504G>T p.M168I Chr.1: 63533471-63533471 on Build GRCh38
4009324 c.568C>T p.R190C Chr.1: 63533535-63533535 on Build GRCh38
293816 c.569G>A p.R190H Chr.1: 63533536-63533536 on Build GRCh38
6440209 c.592G>A p.E198K Chr.1: 63533559-63533559 on Build GRCh38
4571555 c.489T>C p.F163F Chr.1: 63533456-63533456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774502571 Chr.1: 63533428-63533428 on Build GRCh38
rs750941504 Chr.1: 63533535-63533535 on Build GRCh38
rs774992341 Chr.1: 63533438-63533438 on Build GRCh38
rs202119910 Chr.1: 63533426-63533426 on Build GRCh38
rs201173763 Chr.1: 63533443-63533443 on Build GRCh38
rs775685170 Chr.1: 63533476-63533476 on Build GRCh38
rs763322600 Chr.1: 63533415-63533415 on Build GRCh38
rs755134606 Chr.1: 63533554-63533554 on Build GRCh38
rs139429930 Chr.1: 63533563-63533563 on Build GRCh38
rs996939044 Chr.1: 63533378-63533378 on Build GRCh38
rs775289430 Chr.1: 63533410-63533410 on Build GRCh38
rs766476295 Chr.1: 63533544-63533544 on Build GRCh38
rs12041368 Chr.1: 63533552-63533552 on Build GRCh38
rs751991888 Chr.1: 63533420-63533420 on Build GRCh38
rs772447948 Chr.1: 63533588-63533588 on Build GRCh38
rs777402061 Chr.1: 63533446-63533446 on Build GRCh38
rs1027285726 Chr.1: 63533493-63533493 on Build GRCh38
rs217454 Chr.1: 63533416-63533416 on Build GRCh38
rs750494315 Chr.1: 63533438-63533438 on Build GRCh38
rs781221854 Chr.1: 63533559-63533559 on Build GRCh38
rs770760896 Chr.1: 63533453-63533453 on Build GRCh38
rs772663631 Chr.1: 63533469-63533469 on Build GRCh38
rs562743016 Chr.1: 63533521-63533521 on Build GRCh38
rs534803753 Chr.1: 63533577-63533577 on Build GRCh38
rs762547387 Chr.1: 63533501-63533501 on Build GRCh38
rs61738366 Chr.1: 63533516-63533516 on Build GRCh38
rs760321739 Chr.1: 63533437-63533438 on Build GRCh38
rs372824134 Chr.1: 63533536-63533536 on Build GRCh38
rs186494386 Chr.1: 63533432-63533432 on Build GRCh38
rs775237667 Chr.1: 63533481-63533481 on Build GRCh38
rs139108325 Chr.1: 63533480-63533480 on Build GRCh38
rs376080647 Chr.1: 63533556-63533556 on Build GRCh38
rs769391641 Chr.1: 63533569-63533569 on Build GRCh38
rs146388187 Chr.1: 63533558-63533558 on Build GRCh38
rs368888591 Chr.1: 63533557-63533557 on Build GRCh38
rs768401511 Chr.1: 63533508-63533508 on Build GRCh38
rs200199626 Chr.1: 63533452-63533452 on Build GRCh38
rs373427100 Chr.1: 63533409-63533409 on Build GRCh38
rs758170306 Chr.1: 63533444-63533444 on Build GRCh38
rs9436246 Chr.1: 63533523-63533523 on Build GRCh38
rs147278933 Chr.1: 63533467-63533467 on Build GRCh38
rs950206374 Chr.1: 63533541-63533541 on Build GRCh38
rs145293247 Chr.1: 63533587-63533587 on Build GRCh38
rs753551767 Chr.1: 63533578-63533578 on Build GRCh38
rs1054828436 Chr.1: 63533560-63533560 on Build GRCh38

View Full Product Details