Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224481753-224482020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
UniGene
Hs.372286
Gene Chromosome Location
Chr.2: 224470150-224585397 on Build GRCh38
Transcripts
NM_001257197.1 NM_001257198.1 NM_003590.4 XM_006712800.2 XM_011511994.2 XM_011511995.1 XM_011511996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1017109 c.1994T>A p.F665Y Chr.2: 224481927-224481927 on Build GRCh38
6948794 c.1969C>T p.H657Y Chr.2: 224481952-224481952 on Build GRCh38
1692064 c.1939G>A p.E647K Chr.2: 224481982-224481982 on Build GRCh38
6914609 c.2013_2014delAG p.R671fs*12 Chr.2: 224481907-224481908 on Build GRCh38
442398 c.2016C>G p.V672V Chr.2: 224481905-224481905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762108486 Chr.2: 224481846-224481846 on Build GRCh38
rs371959500 Chr.2: 224481879-224481879 on Build GRCh38
rs1040008469 Chr.2: 224481829-224481829 on Build GRCh38
rs773674105 Chr.2: 224481853-224481853 on Build GRCh38
rs553842071 Chr.2: 224481842-224481842 on Build GRCh38
rs1031155134 Chr.2: 224481800-224481800 on Build GRCh38
rs56264124 Chr.2: 224481840-224481840 on Build GRCh38
rs766853487 Chr.2: 224481856-224481856 on Build GRCh38
rs754287382 Chr.2: 224481860-224481860 on Build GRCh38
rs989544747 Chr.2: 224481833-224481833 on Build GRCh38
rs746745736 Chr.2: 224481955-224481955 on Build GRCh38
rs557785257 Chr.2: 224481792-224481792 on Build GRCh38
rs777545905 Chr.2: 224481954-224481954 on Build GRCh38
rs373841352 Chr.2: 224481855-224481855 on Build GRCh38
rs56324678 Chr.2: 224481841-224481841 on Build GRCh38
rs10203789 Chr.2: 224481823-224481823 on Build GRCh38
rs887208304 Chr.2: 224481888-224481888 on Build GRCh38
rs577970903 Chr.2: 224481793-224481793 on Build GRCh38
rs768018900 Chr.2: 224481847-224481847 on Build GRCh38
rs570375246 Chr.2: 224481788-224481788 on Build GRCh38
rs2070127 Chr.2: 224481929-224481929 on Build GRCh38
rs756984902 Chr.2: 224481968-224481968 on Build GRCh38
rs758003374 Chr.2: 224481948-224481948 on Build GRCh38
rs201363693 Chr.2: 224481878-224481878 on Build GRCh38
rs781253532 Chr.2: 224481986-224481986 on Build GRCh38
rs4674912 Chr.2: 224481851-224481851 on Build GRCh38
rs769139389 Chr.2: 224481870-224481870 on Build GRCh38

View Full Product Details