Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18001472-18001697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
UniGene
Hs.515249
Gene Chromosome Location
Chr.19: 18001132-18014102 on Build GRCh38
Transcripts
NM_001025604.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632201 c.250C>T p.L84L Chr.19: 18001564-18001564 on Build GRCh38
6398917 c.165C>A p.H55Q Chr.19: 18001479-18001479 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749370570 Chr.19: 18001628-18001628 on Build GRCh38
rs906585839 Chr.19: 18001655-18001655 on Build GRCh38
rs758798478 Chr.19: 18001611-18001612 on Build GRCh38
rs182672828 Chr.19: 18001560-18001560 on Build GRCh38
rs1028718813 Chr.19: 18001556-18001556 on Build GRCh38
rs939444731 Chr.19: 18001669-18001669 on Build GRCh38
rs776732726 Chr.19: 18001490-18001490 on Build GRCh38
rs987116358 Chr.19: 18001563-18001563 on Build GRCh38
rs746905627 Chr.19: 18001650-18001650 on Build GRCh38
rs748707236 Chr.19: 18001560-18001562 on Build GRCh38
rs1045225026 Chr.19: 18001645-18001645 on Build GRCh38
rs532413635 Chr.19: 18001550-18001550 on Build GRCh38
rs567480721 Chr.19: 18001667-18001667 on Build GRCh38
rs533797268 Chr.19: 18001589-18001589 on Build GRCh38
rs1052564355 Chr.19: 18001673-18001673 on Build GRCh38
rs755580456 Chr.19: 18001615-18001615 on Build GRCh38
rs759759107 Chr.19: 18001541-18001541 on Build GRCh38
rs1021169029 Chr.19: 18001571-18001571 on Build GRCh38
rs907542309 Chr.19: 18001570-18001570 on Build GRCh38
rs1052024081 Chr.19: 18001499-18001499 on Build GRCh38
rs536042256 Chr.19: 18001668-18001668 on Build GRCh38
rs1021583230 Chr.19: 18001508-18001508 on Build GRCh38
rs888775737 Chr.19: 18001503-18001505 on Build GRCh38
rs767498570 Chr.19: 18001577-18001577 on Build GRCh38
rs926583361 Chr.19: 18001625-18001625 on Build GRCh38
rs113540008 Chr.19: 18001552-18001552 on Build GRCh38
rs187802526 Chr.19: 18001624-18001624 on Build GRCh38
rs948195206 Chr.19: 18001641-18001641 on Build GRCh38

View Full Product Details