Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32101174-32101447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA7
Entrez Gene ID
1139
Gene Name
cholinergic receptor nicotinic alpha 7 subunit
Gene Aliases
CHRNA7-2, NACHRA7
UniGene
Hs.511772
Gene Chromosome Location
Chr.15: 32030462-32172521 on Build GRCh38
Transcripts
NM_000746.5 NM_001190455.2 NR_046324.1 XM_011521176.2 XM_011521178.2 XM_017021882.1 XM_017021883.1 XM_017021884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214400 c.283-4G>T p.? Chr.15: 32101299-32101299 on Build GRCh38
6216592 c.240+10G>C p.? Chr.15: 32101357-32101357 on Build GRCh38
3746378 c.196-4G>T p.? Chr.15: 32101299-32101299 on Build GRCh38
6216591 c.327+10G>C p.? Chr.15: 32101357-32101357 on Build GRCh38
6076891 c.234G>T p.L78L Chr.15: 32101341-32101341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749770317 Chr.15: 32101282-32101282 on Build GRCh38
rs1010753983 Chr.15: 32101208-32101209 on Build GRCh38
rs960170256 Chr.15: 32101201-32101201 on Build GRCh38
rs1003839401 Chr.15: 32101200-32101202 on Build GRCh38
rs750529424 Chr.15: 32101261-32101261 on Build GRCh38
rs746629516 Chr.15: 32101283-32101283 on Build GRCh38
rs911909814 Chr.15: 32101249-32101249 on Build GRCh38
rs749611980 Chr.15: 32101347-32101347 on Build GRCh38
rs973216284 Chr.15: 32101395-32101395 on Build GRCh38
rs555153366 Chr.15: 32101280-32101280 on Build GRCh38
rs748577551 Chr.15: 32101285-32101286 on Build GRCh38
rs770248174 Chr.15: 32101287-32101288 on Build GRCh38
rs375004830 Chr.15: 32101262-32101262 on Build GRCh38
rs369729079 Chr.15: 32101396-32101396 on Build GRCh38
rs200823638 Chr.15: 32101367-32101367 on Build GRCh38
rs1022381198 Chr.15: 32101220-32101220 on Build GRCh38
rs201473594 Chr.15: 32101312-32101312 on Build GRCh38
rs761613157 Chr.15: 32101357-32101357 on Build GRCh38
rs906435546 Chr.15: 32101198-32101198 on Build GRCh38
rs766120402 Chr.15: 32101208-32101208 on Build GRCh38
rs760519088 Chr.15: 32101323-32101323 on Build GRCh38
rs190780440 Chr.15: 32101214-32101214 on Build GRCh38
rs34476605 Chr.15: 32101298-32101299 on Build GRCh38
rs796102845 Chr.15: 32101284-32101285 on Build GRCh38
rs775344421 Chr.15: 32101311-32101311 on Build GRCh38
rs201580040 Chr.15: 32101393-32101393 on Build GRCh38
rs778560964 Chr.15: 32101254-32101254 on Build GRCh38
rs986112229 Chr.15: 32101247-32101247 on Build GRCh38
rs771830954 Chr.15: 32101310-32101310 on Build GRCh38
rs575300184 Chr.15: 32101356-32101356 on Build GRCh38
rs984424845 Chr.15: 32101227-32101227 on Build GRCh38
rs201473334 Chr.15: 32101267-32101267 on Build GRCh38
rs200648754 Chr.15: 32101299-32101299 on Build GRCh38
rs367768493 Chr.15: 32101283-32101284 on Build GRCh38
rs956489449 Chr.15: 32101206-32101206 on Build GRCh38
rs747878364 Chr.15: 32101282-32101283 on Build GRCh38
rs750156618 Chr.15: 32101284-32101284 on Build GRCh38
rs867466139 Chr.15: 32101301-32101301 on Build GRCh38
rs763734880 Chr.15: 32101348-32101348 on Build GRCh38
rs748546932 Chr.15: 32101282-32101282 on Build GRCh38
rs751665278 Chr.15: 32101397-32101397 on Build GRCh38
rs755363492 Chr.15: 32101276-32101276 on Build GRCh38
rs745413698 Chr.15: 32101300-32101300 on Build GRCh38
rs773457639 Chr.15: 32101299-32101318 on Build GRCh38
rs953481760 Chr.15: 32101239-32101239 on Build GRCh38
rs200083680 Chr.15: 32101326-32101326 on Build GRCh38
rs200305897 Chr.15: 32101373-32101373 on Build GRCh38
rs970293711 Chr.15: 32101226-32101226 on Build GRCh38
rs780301216 Chr.15: 32101266-32101266 on Build GRCh38
rs12899798 Chr.15: 32101336-32101336 on Build GRCh38
rs201136574 Chr.15: 32101380-32101380 on Build GRCh38
rs750478145 Chr.15: 32101371-32101371 on Build GRCh38
rs758530751 Chr.15: 32101379-32101379 on Build GRCh38
rs199797163 Chr.15: 32101283-32101283 on Build GRCh38

View Full Product Details