Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23451769-23452042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST11
Entrez Gene ID
140880
Gene Name
cystatin 11
Gene Aliases
CST8L, CTES2, SC13, dJ322G13.6
UniGene
Hs.128100
Gene Chromosome Location
Chr.20: 23450404-23452845 on Build GRCh38
Transcripts
NM_080830.2 NM_130794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280191 c.300G>A p.T100T Chr.20: 23451849-23451849 on Build GRCh38
1025223 c.253C>A p.L85M Chr.20: 23451896-23451896 on Build GRCh38
319722 c.237C>A p.D79E Chr.20: 23451912-23451912 on Build GRCh38
723349 c.325C>T p.L109F Chr.20: 23451824-23451824 on Build GRCh38
6159607 c.312C>A p.P104P Chr.20: 23451837-23451837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747371325 Chr.20: 23451821-23451821 on Build GRCh38
rs372121340 Chr.20: 23451805-23451805 on Build GRCh38
rs889838116 Chr.20: 23451796-23451796 on Build GRCh38
rs770430472 Chr.20: 23451849-23451849 on Build GRCh38
rs771385100 Chr.20: 23451824-23451824 on Build GRCh38
rs944083826 Chr.20: 23451956-23451956 on Build GRCh38
rs748747604 Chr.20: 23451818-23451818 on Build GRCh38
rs781075575 Chr.20: 23451794-23451794 on Build GRCh38
rs143205954 Chr.20: 23451842-23451842 on Build GRCh38
rs750229124 Chr.20: 23451880-23451880 on Build GRCh38
rs764289870 Chr.20: 23451793-23451793 on Build GRCh38
rs777145660 Chr.20: 23452023-23452023 on Build GRCh38
rs143202878 Chr.20: 23451820-23451820 on Build GRCh38
rs758120186 Chr.20: 23451969-23451969 on Build GRCh38
rs750013735 Chr.20: 23451924-23451924 on Build GRCh38
rs36010049 Chr.20: 23451897-23451897 on Build GRCh38
rs201144078 Chr.20: 23451928-23451928 on Build GRCh38
rs371350578 Chr.20: 23451884-23451884 on Build GRCh38
rs770593668 Chr.20: 23452004-23452004 on Build GRCh38
rs758138699 Chr.20: 23451921-23451921 on Build GRCh38
rs775910071 Chr.20: 23451932-23451932 on Build GRCh38
rs6137952 Chr.20: 23451870-23451870 on Build GRCh38
rs141906805 Chr.20: 23452006-23452006 on Build GRCh38
rs999911545 Chr.20: 23451985-23451985 on Build GRCh38
rs914266924 Chr.20: 23451828-23451828 on Build GRCh38
rs6114090 Chr.20: 23451997-23451997 on Build GRCh38
rs183565619 Chr.20: 23451858-23451858 on Build GRCh38
rs375509708 Chr.20: 23451935-23451935 on Build GRCh38
rs1023349534 Chr.20: 23451862-23451862 on Build GRCh38
rs764596521 Chr.20: 23451965-23451965 on Build GRCh38
rs770410426 Chr.20: 23451930-23451930 on Build GRCh38
rs552714338 Chr.20: 23451868-23451868 on Build GRCh38
rs542052853 Chr.20: 23451938-23451938 on Build GRCh38
rs920133009 Chr.20: 23451926-23451926 on Build GRCh38
rs1032264696 Chr.20: 23451996-23451996 on Build GRCh38
rs755996261 Chr.20: 23451905-23451906 on Build GRCh38
rs908074895 Chr.20: 23452012-23452012 on Build GRCh38
rs767248934 Chr.20: 23451871-23451871 on Build GRCh38
rs1019897610 Chr.20: 23451958-23451958 on Build GRCh38
rs970475315 Chr.20: 23452007-23452007 on Build GRCh38
rs764354728 Chr.20: 23451865-23451865 on Build GRCh38
rs777159188 Chr.20: 23451838-23451838 on Build GRCh38
rs375491260 Chr.20: 23451850-23451850 on Build GRCh38
rs754967899 Chr.20: 23451950-23451950 on Build GRCh38
rs376419554 Chr.20: 23451970-23451970 on Build GRCh38
rs6114091 Chr.20: 23452013-23452013 on Build GRCh38
rs537611679 Chr.20: 23451974-23451974 on Build GRCh38
rs953918347 Chr.20: 23451802-23451802 on Build GRCh38
rs6114089 Chr.20: 23451967-23451967 on Build GRCh38
rs545515085 Chr.20: 23451819-23451819 on Build GRCh38
rs779478256 Chr.20: 23451806-23451806 on Build GRCh38
rs777642106 Chr.20: 23451909-23451915 on Build GRCh38
rs369693661 Chr.20: 23451998-23451998 on Build GRCh38
rs568017076 Chr.20: 23451890-23451890 on Build GRCh38
rs576847119 Chr.20: 23451936-23451936 on Build GRCh38
rs753165979 Chr.20: 23451975-23451975 on Build GRCh38
rs148274683 Chr.20: 23451904-23451904 on Build GRCh38
rs747708496 Chr.20: 23451919-23451919 on Build GRCh38
rs748765306 Chr.20: 23451924-23451925 on Build GRCh38
rs531397640 Chr.20: 23451867-23451867 on Build GRCh38
rs555927756 Chr.20: 23451940-23451940 on Build GRCh38
rs1028250261 Chr.20: 23451937-23451937 on Build GRCh38
rs981783251 Chr.20: 23451878-23451878 on Build GRCh38
rs980140953 Chr.20: 23451977-23451977 on Build GRCh38
rs200225444 Chr.20: 23451929-23451929 on Build GRCh38
rs766275966 Chr.20: 23451939-23451939 on Build GRCh38
rs2236021 Chr.20: 23451891-23451891 on Build GRCh38

View Full Product Details