Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46397504-46397773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
UniGene
Hs.654644
Gene Chromosome Location
Chr.11: 46396412-46594069 on Build GRCh38
Transcripts
NM_001267782.1 NM_001267783.1 NM_001300731.1 NM_017749.3 XM_005253009.3 XM_005253011.3 XM_005253014.3 XM_006718259.2 XM_006718260.2 XM_017018007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3447583 c.3823A>G p.N1275D Chr.11: 46397524-46397524 on Build GRCh38
429009 c.3691G>C p.A1231P Chr.11: 46397656-46397656 on Build GRCh38
189529 c.3311C>T p.T1104M Chr.11: 46397766-46397766 on Build GRCh38
299857 c.3450delG p.R1151fs*32 Chr.11: 46397627-46397627 on Build GRCh38
7114833 c.3762C>G p.V1254V Chr.11: 46397585-46397585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779975161 Chr.11: 46397729-46397729 on Build GRCh38
rs374289799 Chr.11: 46397600-46397600 on Build GRCh38
rs762197243 Chr.11: 46397661-46397661 on Build GRCh38
rs769004853 Chr.11: 46397742-46397742 on Build GRCh38
rs553288239 Chr.11: 46397726-46397726 on Build GRCh38
rs377430582 Chr.11: 46397692-46397692 on Build GRCh38
rs577271663 Chr.11: 46397747-46397747 on Build GRCh38
rs921662645 Chr.11: 46397575-46397575 on Build GRCh38
rs748009899 Chr.11: 46397606-46397606 on Build GRCh38
rs200914758 Chr.11: 46397607-46397607 on Build GRCh38
rs781305322 Chr.11: 46397695-46397695 on Build GRCh38
rs754709228 Chr.11: 46397572-46397572 on Build GRCh38
rs776949386 Chr.11: 46397657-46397657 on Build GRCh38
rs144388436 Chr.11: 46397746-46397746 on Build GRCh38
rs764821864 Chr.11: 46397673-46397673 on Build GRCh38
rs142425575 Chr.11: 46397662-46397662 on Build GRCh38
rs752488700 Chr.11: 46397696-46397696 on Build GRCh38
rs866037672 Chr.11: 46397748-46397748 on Build GRCh38
rs149624973 Chr.11: 46397712-46397712 on Build GRCh38
rs964199125 Chr.11: 46397560-46397560 on Build GRCh38
rs145195351 Chr.11: 46397537-46397537 on Build GRCh38
rs745882997 Chr.11: 46397616-46397616 on Build GRCh38
rs759715949 Chr.11: 46397530-46397530 on Build GRCh38
rs200841087 Chr.11: 46397713-46397713 on Build GRCh38
rs375911943 Chr.11: 46397691-46397691 on Build GRCh38
rs749895755 Chr.11: 46397596-46397596 on Build GRCh38
rs769253896 Chr.11: 46397656-46397656 on Build GRCh38
rs770151053 Chr.11: 46397749-46397749 on Build GRCh38
rs138262595 Chr.11: 46397693-46397693 on Build GRCh38
rs747977891 Chr.11: 46397594-46397594 on Build GRCh38
rs775703311 Chr.11: 46397631-46397631 on Build GRCh38
rs1023861260 Chr.11: 46397677-46397677 on Build GRCh38
rs866207238 Chr.11: 46397577-46397577 on Build GRCh38
rs766988577 Chr.11: 46397733-46397735 on Build GRCh38
rs764361329 Chr.11: 46397547-46397547 on Build GRCh38
rs762460695 Chr.11: 46397686-46397686 on Build GRCh38
rs751458401 Chr.11: 46397602-46397602 on Build GRCh38
rs768017118 Chr.11: 46397533-46397533 on Build GRCh38
rs771340316 Chr.11: 46397528-46397528 on Build GRCh38
rs780837593 Chr.11: 46397582-46397582 on Build GRCh38
rs754257749 Chr.11: 46397558-46397558 on Build GRCh38
rs761032235 Chr.11: 46397546-46397546 on Build GRCh38
rs761245404 Chr.11: 46397600-46397601 on Build GRCh38
rs761071769 Chr.11: 46397642-46397642 on Build GRCh38
rs370167428 Chr.11: 46397709-46397709 on Build GRCh38
rs747152677 Chr.11: 46397741-46397741 on Build GRCh38
rs137890582 Chr.11: 46397603-46397603 on Build GRCh38
rs61733783 Chr.11: 46397576-46397576 on Build GRCh38
rs779296190 Chr.11: 46397610-46397610 on Build GRCh38
rs554226184 Chr.11: 46397697-46397697 on Build GRCh38
rs772046544 Chr.11: 46397626-46397626 on Build GRCh38
rs117438791 Chr.11: 46397676-46397676 on Build GRCh38
rs374387714 Chr.11: 46397755-46397755 on Build GRCh38

View Full Product Details