Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50351916-50352151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561D2
Entrez Gene ID
11068
Gene Name
cytochrome b561 family member D2
Gene Aliases
101F6, TSP10, XXcos-LUCA11.4
UniGene
Hs.149443
Gene Chromosome Location
Chr.3: 50350695-50368197 on Build GRCh38
Transcripts
NM_001291284.1 NM_007022.4 NR_111912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2851378 c.138C>A p.S46S Chr.3: 50352019-50352019 on Build GRCh38
48309 c.142C>A p.H48N Chr.3: 50352023-50352023 on Build GRCh38
1046510 c.135C>T p.F45F Chr.3: 50352016-50352016 on Build GRCh38
6468979 c.165T>C p.A55A Chr.3: 50352046-50352046 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764073654 Chr.3: 50351992-50351992 on Build GRCh38
rs767698734 Chr.3: 50352065-50352065 on Build GRCh38
rs767643494 Chr.3: 50351981-50351981 on Build GRCh38
rs200062155 Chr.3: 50351989-50351989 on Build GRCh38
rs145727263 Chr.3: 50352028-50352028 on Build GRCh38
rs141123544 Chr.3: 50352007-50352007 on Build GRCh38
rs779776068 Chr.3: 50352020-50352020 on Build GRCh38
rs759823974 Chr.3: 50352063-50352063 on Build GRCh38
rs587706761 Chr.3: 50352112-50352112 on Build GRCh38
rs759697135 Chr.3: 50351979-50351979 on Build GRCh38
rs771708334 Chr.3: 50351965-50351965 on Build GRCh38
rs752821236 Chr.3: 50351984-50351984 on Build GRCh38
rs773488280 Chr.3: 50351972-50351972 on Build GRCh38
rs140638768 Chr.3: 50351955-50351956 on Build GRCh38
rs1007867837 Chr.3: 50351955-50351955 on Build GRCh38
rs762536149 Chr.3: 50352016-50352018 on Build GRCh38
rs757147135 Chr.3: 50352010-50352010 on Build GRCh38
rs370367265 Chr.3: 50352057-50352057 on Build GRCh38
rs753672499 Chr.3: 50351996-50351996 on Build GRCh38
rs745525518 Chr.3: 50352018-50352018 on Build GRCh38
rs947441650 Chr.3: 50352106-50352106 on Build GRCh38
rs761781659 Chr.3: 50352095-50352095 on Build GRCh38
rs766691148 Chr.3: 50351974-50351974 on Build GRCh38
rs775078351 Chr.3: 50351967-50351967 on Build GRCh38
rs770251945 Chr.3: 50352027-50352027 on Build GRCh38
rs969206945 Chr.3: 50352035-50352035 on Build GRCh38
rs776715489 Chr.3: 50352021-50352021 on Build GRCh38
rs1057226252 Chr.3: 50352075-50352075 on Build GRCh38
rs200349019 Chr.3: 50351964-50351964 on Build GRCh38
rs148275530 Chr.3: 50352066-50352066 on Build GRCh38
rs372358872 Chr.3: 50352071-50352071 on Build GRCh38
rs774563388 Chr.3: 50351978-50351978 on Build GRCh38
rs750651635 Chr.3: 50351990-50351992 on Build GRCh38
rs758107728 Chr.3: 50352019-50352019 on Build GRCh38
rs373812398 Chr.3: 50352059-50352059 on Build GRCh38
rs770329940 Chr.3: 50351970-50351970 on Build GRCh38
rs192405831 Chr.3: 50352092-50352092 on Build GRCh38
rs774877638 Chr.3: 50352094-50352094 on Build GRCh38
rs75420946 Chr.3: 50352046-50352046 on Build GRCh38
rs909031209 Chr.3: 50352055-50352055 on Build GRCh38
rs1050549182 Chr.3: 50352085-50352085 on Build GRCh38
rs763353262 Chr.3: 50351973-50351973 on Build GRCh38
rs772002927 Chr.3: 50352013-50352013 on Build GRCh38
rs765863754 Chr.3: 50352031-50352031 on Build GRCh38

View Full Product Details