Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20807106-20807377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
UniGene
Hs.467084
Gene Chromosome Location
Chr.1: 20806292-21177320 on Build GRCh38
Transcripts
NM_001198801.1 NM_001198802.1 NM_003760.4 XM_011542356.2 XM_011542361.2 XM_011542370.1 XM_011542371.1 XM_011542374.1 XM_011542375.2 XM_011542381.2 XM_017002672.1 XM_017002673.1 XM_017002674.1 XM_017002675.1 XM_017002676.1 XM_017002677.1 XM_017002678.1 XM_017002679.1 XM_017002680.1 XM_017002681.1 XM_017002682.1 XM_017002683.1 XM_017002684.1 XM_017002685.1 XM_017002686.1 XM_017002687.1 XM_017002688.1 XM_017002689.1 XM_017002690.1 XM_017002691.1 XM_017002692.1 XM_017002693.1 XM_017002694.1 XM_017002695.1 XM_017002696.1 XM_017002697.1 XM_017002698.1 XM_017002699.1 XM_017002700.1 XM_017002701.1 XM_017002702.1 XM_017002703.1 XM_017002704.1 XM_017002705.1 XM_017002706.1 XM_017002707.1 XM_017002708.1 XM_017002709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653028 c.4719G>A p.T1573T Chr.1: 20807358-20807358 on Build GRCh38
5590741 c.3170C>T p.S1057F Chr.1: 20807377-20807377 on Build GRCh38
7136417 c.4722C>A p.V1574V Chr.1: 20807373-20807373 on Build GRCh38
3966239 c.4708G>A p.A1570T Chr.1: 20807369-20807369 on Build GRCh38
6540169 c.3219G>A p.E1073E Chr.1: 20807328-20807328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762166851 Chr.1: 20807299-20807299 on Build GRCh38
rs1006167053 Chr.1: 20807172-20807172 on Build GRCh38
rs371811192 Chr.1: 20807210-20807210 on Build GRCh38
rs754443566 Chr.1: 20807307-20807307 on Build GRCh38
rs146669269 Chr.1: 20807343-20807343 on Build GRCh38
rs754223109 Chr.1: 20807286-20807286 on Build GRCh38
rs907719117 Chr.1: 20807207-20807207 on Build GRCh38
rs1047148439 Chr.1: 20807194-20807194 on Build GRCh38
rs767740107 Chr.1: 20807268-20807268 on Build GRCh38
rs775476606 Chr.1: 20807269-20807269 on Build GRCh38
rs772621170 Chr.1: 20807291-20807291 on Build GRCh38
rs776091222 Chr.1: 20807308-20807309 on Build GRCh38
rs202173650 Chr.1: 20807303-20807303 on Build GRCh38
rs765956273 Chr.1: 20807300-20807300 on Build GRCh38
rs375666603 Chr.1: 20807220-20807220 on Build GRCh38
rs756586385 Chr.1: 20807229-20807229 on Build GRCh38
rs780721683 Chr.1: 20807309-20807309 on Build GRCh38
rs887678267 Chr.1: 20807172-20807174 on Build GRCh38
rs77692149 Chr.1: 20807178-20807178 on Build GRCh38
rs766973359 Chr.1: 20807202-20807202 on Build GRCh38
rs1045106999 Chr.1: 20807127-20807127 on Build GRCh38
rs761226476 Chr.1: 20807271-20807271 on Build GRCh38
rs979665832 Chr.1: 20807283-20807283 on Build GRCh38
rs756923767 Chr.1: 20807198-20807198 on Build GRCh38
rs746363580 Chr.1: 20807268-20807269 on Build GRCh38
rs764563737 Chr.1: 20807281-20807281 on Build GRCh38
rs931392081 Chr.1: 20807171-20807171 on Build GRCh38
rs752197358 Chr.1: 20807315-20807315 on Build GRCh38
rs1021312049 Chr.1: 20807246-20807246 on Build GRCh38

View Full Product Details